Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 175006997-175007270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACYBP
Entrez Gene ID
27101
Gene Name
calcyclin binding protein
Gene Aliases
GIG5, PNAS-107, S100A6BP, SIP
UniGene
Hs.508524
Gene Chromosome Location
Chr.1: 174999435-175012027 on Build GRCh38
Transcripts
NM_001007214.1 NM_014412.2 XM_017001046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1336409 c.379A>G p.M127V Chr.1: 175007144-175007144 on Build GRCh38
677040 c.381G>A p.M127I Chr.1: 175007146-175007146 on Build GRCh38
1626637 c.344T>G p.L115R Chr.1: 175007109-175007109 on Build GRCh38
5844559 c.415G>A p.E139K Chr.1: 175007180-175007180 on Build GRCh38
7195725 c.333-1G>A p.? Chr.1: 175007097-175007097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs16847689 Chr.1: 175007048-175007048 on Build GRCh38
rs1049809882 Chr.1: 175007169-175007169 on Build GRCh38
rs754817774 Chr.1: 175007071-175007073 on Build GRCh38
rs769306905 Chr.1: 175007221-175007221 on Build GRCh38
rs936689171 Chr.1: 175007147-175007147 on Build GRCh38
rs769991041 Chr.1: 175007208-175007210 on Build GRCh38
rs150909890 Chr.1: 175007128-175007128 on Build GRCh38
rs569818004 Chr.1: 175007137-175007137 on Build GRCh38
rs772710020 Chr.1: 175007217-175007217 on Build GRCh38
rs758990793 Chr.1: 175007220-175007220 on Build GRCh38
rs750475722 Chr.1: 175007139-175007139 on Build GRCh38
rs186009547 Chr.1: 175007235-175007235 on Build GRCh38
rs762764567 Chr.1: 175007064-175007064 on Build GRCh38
rs539831859 Chr.1: 175007111-175007111 on Build GRCh38
rs748470625 Chr.1: 175007207-175007208 on Build GRCh38
rs763911991 Chr.1: 175007067-175007067 on Build GRCh38
rs780786686 Chr.1: 175007082-175007084 on Build GRCh38
rs992048172 Chr.1: 175007134-175007134 on Build GRCh38
rs938170632 Chr.1: 175007027-175007027 on Build GRCh38
rs945607640 Chr.1: 175007052-175007052 on Build GRCh38
rs751330585 Chr.1: 175007070-175007070 on Build GRCh38
rs1042694422 Chr.1: 175007101-175007101 on Build GRCh38
rs369204691 Chr.1: 175007051-175007051 on Build GRCh38
rs748510976 Chr.1: 175007210-175007210 on Build GRCh38
rs548172319 Chr.1: 175007019-175007020 on Build GRCh38
rs757240575 Chr.1: 175007116-175007116 on Build GRCh38
rs762640166 Chr.1: 175007241-175007241 on Build GRCh38
rs774828117 Chr.1: 175007225-175007225 on Build GRCh38
rs756165044 Chr.1: 175007156-175007156 on Build GRCh38
rs749482584 Chr.1: 175007173-175007173 on Build GRCh38
rs761697071 Chr.1: 175007020-175007020 on Build GRCh38
rs201531530 Chr.1: 175007175-175007175 on Build GRCh38
rs763719782 Chr.1: 175007243-175007243 on Build GRCh38
rs992749826 Chr.1: 175007031-175007031 on Build GRCh38
rs780166032 Chr.1: 175007172-175007172 on Build GRCh38
rs200258548 Chr.1: 175007118-175007118 on Build GRCh38

View Full Product Details