Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153951871-153952134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC2
Entrez Gene ID
200186
Gene Name
CREB regulated transcription coactivator 2
Gene Aliases
TORC-2, TORC2
UniGene
Hs.406392
Gene Chromosome Location
Chr.1: 153947672-153958656 on Build GRCh38
Transcripts
NM_181715.2 XM_005244946.1 XM_005244949.2 XM_017000575.1 XM_017000576.1 XM_017000577.1 XM_017000578.1 XM_017000579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2208366 c.918T>C p.S306S Chr.1: 153952097-153952097 on Build GRCh38
7158445 c.988G>C p.D330H Chr.1: 153952027-153952027 on Build GRCh38
7190586 c.955A>T p.I319F Chr.1: 153952060-153952060 on Build GRCh38
4935928 c.996A>G p.P332P Chr.1: 153952019-153952019 on Build GRCh38
3984265 c.971G>T p.G324V Chr.1: 153952044-153952044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767750977 Chr.1: 153952024-153952024 on Build GRCh38
rs911199642 Chr.1: 153952081-153952081 on Build GRCh38
rs972977955 Chr.1: 153951931-153951931 on Build GRCh38
rs760822150 Chr.1: 153952029-153952029 on Build GRCh38
rs764333945 Chr.1: 153952048-153952048 on Build GRCh38
rs763451339 Chr.1: 153952001-153952001 on Build GRCh38
rs747624733 Chr.1: 153951967-153951967 on Build GRCh38
rs753020386 Chr.1: 153952026-153952026 on Build GRCh38
rs746224206 Chr.1: 153952104-153952104 on Build GRCh38
rs568488100 Chr.1: 153951925-153951925 on Build GRCh38
rs765263592 Chr.1: 153951899-153951899 on Build GRCh38
rs751329018 Chr.1: 153952067-153952067 on Build GRCh38
rs990276946 Chr.1: 153951945-153951945 on Build GRCh38
rs879757775 Chr.1: 153951952-153951952 on Build GRCh38
rs769441269 Chr.1: 153952084-153952084 on Build GRCh38
rs745833322 Chr.1: 153951972-153951973 on Build GRCh38
rs773706306 Chr.1: 153951998-153951998 on Build GRCh38
rs367678333 Chr.1: 153951900-153951900 on Build GRCh38
rs758294887 Chr.1: 153952051-153952051 on Build GRCh38
rs547941993 Chr.1: 153951992-153951992 on Build GRCh38
rs779877153 Chr.1: 153952057-153952057 on Build GRCh38
rs149606285 Chr.1: 153951946-153951946 on Build GRCh38
rs780799759 Chr.1: 153952075-153952075 on Build GRCh38
rs916703544 Chr.1: 153952100-153952100 on Build GRCh38
rs777207213 Chr.1: 153952092-153952092 on Build GRCh38
rs542980300 Chr.1: 153952004-153952004 on Build GRCh38
rs961199783 Chr.1: 153951916-153951916 on Build GRCh38
rs759853966 Chr.1: 153952010-153952010 on Build GRCh38
rs200091206 Chr.1: 153952050-153952050 on Build GRCh38
rs751601004 Chr.1: 153952082-153952082 on Build GRCh38
rs369302090 Chr.1: 153952071-153952071 on Build GRCh38
rs566679859 Chr.1: 153951997-153951997 on Build GRCh38
rs966388956 Chr.1: 153951897-153951897 on Build GRCh38
rs373911120 Chr.1: 153952101-153952101 on Build GRCh38
rs748774074 Chr.1: 153952098-153952098 on Build GRCh38
rs112021980 Chr.1: 153952016-153952016 on Build GRCh38
rs771355755 Chr.1: 153952002-153952002 on Build GRCh38

View Full Product Details