Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152652491-152652738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRQ
Entrez Gene ID
55879
Gene Name
gamma-aminobutyric acid type A receptor theta subunit
Gene Aliases
THETA
UniGene
Hs.283081
Gene Chromosome Location
Chr.X: 152637895-152659827 on Build GRCh38
Transcripts
NM_018558.3 XM_011531184.2

SNP Detail

SNP ID Chromosome Location
rs782510679 Chr.X: 152652653-152652653 on Build GRCh38
rs782272412 Chr.X: 152652716-152652716 on Build GRCh38
rs141690321 Chr.X: 152652646-152652646 on Build GRCh38
rs919456266 Chr.X: 152652669-152652669 on Build GRCh38
rs781904427 Chr.X: 152652551-152652551 on Build GRCh38
rs782741890 Chr.X: 152652665-152652665 on Build GRCh38
rs143687531 Chr.X: 152652701-152652701 on Build GRCh38
rs376533105 Chr.X: 152652577-152652577 on Build GRCh38
rs1009596372 Chr.X: 152652546-152652546 on Build GRCh38
rs782168254 Chr.X: 152652635-152652635 on Build GRCh38
rs782573307 Chr.X: 152652525-152652525 on Build GRCh38
rs1033610208 Chr.X: 152652575-152652575 on Build GRCh38
rs782603061 Chr.X: 152652711-152652711 on Build GRCh38
rs782366909 Chr.X: 152652615-152652615 on Build GRCh38
rs782726466 Chr.X: 152652627-152652627 on Build GRCh38
rs79313209 Chr.X: 152652588-152652588 on Build GRCh38
rs782370340 Chr.X: 152652564-152652564 on Build GRCh38
rs782246365 Chr.X: 152652558-152652558 on Build GRCh38
rs374632460 Chr.X: 152652622-152652622 on Build GRCh38
rs782013942 Chr.X: 152652604-152652604 on Build GRCh38
rs889394845 Chr.X: 152652706-152652706 on Build GRCh38
rs141027732 Chr.X: 152652712-152652712 on Build GRCh38
rs933495335 Chr.X: 152652675-152652675 on Build GRCh38
rs782330878 Chr.X: 152652605-152652605 on Build GRCh38
rs77878508 Chr.X: 152652719-152652719 on Build GRCh38
rs781920686 Chr.X: 152652630-152652630 on Build GRCh38
rs970585130 Chr.X: 152652557-152652557 on Build GRCh38
rs782079877 Chr.X: 152652625-152652625 on Build GRCh38
rs781958098 Chr.X: 152652706-152652707 on Build GRCh38
rs200635320 Chr.X: 152652703-152652703 on Build GRCh38
rs782799189 Chr.X: 152652638-152652638 on Build GRCh38
rs782176060 Chr.X: 152652584-152652584 on Build GRCh38

View Full Product Details