Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 33532907-33533176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSMD2
Entrez Gene ID
114784
Gene Name
CUB and Sushi multiple domains 2
Gene Aliases
dJ1007G16.1, dJ1007G16.2, dJ947L8.1
UniGene
Hs.656915
Gene Chromosome Location
Chr.1: 33513999-34165842 on Build GRCh38
Transcripts
NM_001281956.1 NM_052896.4 XM_017000185.1 XM_017000186.1 XM_017000187.1 XM_017000188.1 XM_017000189.1 XM_017000190.1 XM_017000194.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1661288 c.9722A>T p.K3241M Chr.1: 33533067-33533067 on Build GRCh38
5380700 c.9613G>A p.D3205N Chr.1: 33533176-33533176 on Build GRCh38
5380701 c.10045G>A p.D3349N Chr.1: 33533176-33533176 on Build GRCh38
5990858 c.9674delG p.G3225fs*43 Chr.1: 33533115-33533115 on Build GRCh38
2078991 c.9711C>G p.S3237R Chr.1: 33533078-33533078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752097807 Chr.1: 33532995-33532995 on Build GRCh38
rs767008079 Chr.1: 33533150-33533150 on Build GRCh38
rs564708351 Chr.1: 33533053-33533053 on Build GRCh38
rs950788542 Chr.1: 33532955-33532955 on Build GRCh38
rs1044263131 Chr.1: 33533034-33533034 on Build GRCh38
rs140945113 Chr.1: 33533111-33533111 on Build GRCh38
rs553649805 Chr.1: 33532944-33532944 on Build GRCh38
rs146860302 Chr.1: 33533063-33533063 on Build GRCh38
rs777036386 Chr.1: 33533066-33533066 on Build GRCh38
rs756003035 Chr.1: 33533022-33533022 on Build GRCh38
rs891390487 Chr.1: 33533107-33533107 on Build GRCh38
rs368016507 Chr.1: 33533011-33533011 on Build GRCh38
rs777416603 Chr.1: 33533032-33533032 on Build GRCh38
rs1040353324 Chr.1: 33533132-33533132 on Build GRCh38
rs201781858 Chr.1: 33533147-33533147 on Build GRCh38
rs1027763070 Chr.1: 33532953-33532953 on Build GRCh38
rs561566800 Chr.1: 33533133-33533133 on Build GRCh38
rs368196378 Chr.1: 33533074-33533074 on Build GRCh38
rs561053865 Chr.1: 33533013-33533013 on Build GRCh38
rs754519907 Chr.1: 33533129-33533129 on Build GRCh38
rs372068474 Chr.1: 33533108-33533108 on Build GRCh38
rs544963998 Chr.1: 33533088-33533088 on Build GRCh38
rs371916613 Chr.1: 33533031-33533031 on Build GRCh38
rs755663747 Chr.1: 33533154-33533154 on Build GRCh38
rs770707605 Chr.1: 33533044-33533044 on Build GRCh38
rs149247781 Chr.1: 33532988-33532988 on Build GRCh38
rs973643190 Chr.1: 33532952-33532952 on Build GRCh38
rs267598562 Chr.1: 33533142-33533142 on Build GRCh38
rs542401404 Chr.1: 33533007-33533007 on Build GRCh38
rs747765212 Chr.1: 33533018-33533018 on Build GRCh38
rs749261958 Chr.1: 33533033-33533033 on Build GRCh38
rs773396266 Chr.1: 33533101-33533101 on Build GRCh38
rs144512012 Chr.1: 33533087-33533087 on Build GRCh38
rs775744927 Chr.1: 33533050-33533050 on Build GRCh38
rs541737634 Chr.1: 33532933-33532933 on Build GRCh38
rs1007172601 Chr.1: 33533131-33533131 on Build GRCh38
rs372500958 Chr.1: 33533015-33533015 on Build GRCh38
rs779216459 Chr.1: 33532934-33532934 on Build GRCh38
rs752036680 Chr.1: 33533118-33533118 on Build GRCh38
rs370899719 Chr.1: 33533028-33533037 on Build GRCh38
rs758821750 Chr.1: 33533010-33533010 on Build GRCh38
rs376654604 Chr.1: 33533153-33533153 on Build GRCh38
rs200333295 Chr.1: 33533016-33533016 on Build GRCh38

View Full Product Details