Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 56949386-56949635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8B
Entrez Gene ID
732
Gene Name
complement component 8, beta polypeptide
Gene Aliases
C82
UniGene
Hs.391835
Gene Chromosome Location
Chr.1: 56929210-56974383 on Build GRCh38
Transcripts
NM_000066.3 NM_001278543.1 NM_001278544.1 XM_017002235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7294708 c.850C>T p.R284* Chr.1: 56949569-56949569 on Build GRCh38
3491211 c.860A>G p.H287R Chr.1: 56949559-56949559 on Build GRCh38
239092 c.815G>A p.S272N Chr.1: 56949604-56949604 on Build GRCh38
3805643 c.793G>A p.E265K Chr.1: 56949626-56949626 on Build GRCh38
4559147 c.793G>C p.E265Q Chr.1: 56949626-56949626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769763661 Chr.1: 56949579-56949579 on Build GRCh38
rs148116725 Chr.1: 56949464-56949464 on Build GRCh38
rs112534346 Chr.1: 56949473-56949473 on Build GRCh38
rs113144988 Chr.1: 56949528-56949528 on Build GRCh38
rs751007726 Chr.1: 56949525-56949525 on Build GRCh38
rs1010975053 Chr.1: 56949453-56949453 on Build GRCh38
rs762754727 Chr.1: 56949582-56949582 on Build GRCh38
rs202207968 Chr.1: 56949568-56949568 on Build GRCh38
rs779217317 Chr.1: 56949604-56949604 on Build GRCh38
rs706481 Chr.1: 56949527-56949527 on Build GRCh38
rs1054982999 Chr.1: 56949432-56949432 on Build GRCh38
rs897194268 Chr.1: 56949433-56949433 on Build GRCh38
rs553393814 Chr.1: 56949581-56949581 on Build GRCh38
rs746431472 Chr.1: 56949539-56949540 on Build GRCh38
rs539779449 Chr.1: 56949496-56949496 on Build GRCh38
rs756647598 Chr.1: 56949596-56949596 on Build GRCh38
rs776093033 Chr.1: 56949573-56949573 on Build GRCh38
rs1038658230 Chr.1: 56949408-56949408 on Build GRCh38
rs893611213 Chr.1: 56949449-56949449 on Build GRCh38
rs199765748 Chr.1: 56949584-56949584 on Build GRCh38
rs142497373 Chr.1: 56949600-56949600 on Build GRCh38
rs777354169 Chr.1: 56949551-56949551 on Build GRCh38
rs780551849 Chr.1: 56949598-56949598 on Build GRCh38
rs771635483 Chr.1: 56949548-56949548 on Build GRCh38
rs147387692 Chr.1: 56949590-56949590 on Build GRCh38
rs367587572 Chr.1: 56949586-56949586 on Build GRCh38
rs112330011 Chr.1: 56949420-56949420 on Build GRCh38
rs374155702 Chr.1: 56949569-56949569 on Build GRCh38
rs781773309 Chr.1: 56949510-56949510 on Build GRCh38
rs759525387 Chr.1: 56949577-56949577 on Build GRCh38
rs140813121 Chr.1: 56949599-56949599 on Build GRCh38
rs573966461 Chr.1: 56949483-56949483 on Build GRCh38
rs968587292 Chr.1: 56949550-56949550 on Build GRCh38

View Full Product Details