Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31642237-31642462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG6
Entrez Gene ID
7917
Gene Name
BCL2 associated athanogene 6
Gene Aliases
BAG-6, BAT3, D6S52E, G3
UniGene
Hs.440900
Gene Chromosome Location
Chr.6: 31639028-31660900 on Build GRCh38
Transcripts
NM_001098534.1 NM_001199697.1 NM_001199698.1 NM_004639.3 NM_080702.2 NM_080703.2 XM_011514892.2 XM_017011275.1 XM_017011276.1 XM_017011277.1 XM_017011278.1 XM_017011279.1 XM_017011280.1 XM_017011281.1 XM_017011282.1 XM_017011283.1 XM_017011284.1 XM_017011285.1 XM_017011286.1 XM_017011287.1 XM_017011288.1 XM_017011289.1 XM_017011290.1 XM_017011291.1 XM_017011292.1 XM_017011293.1 XM_017011294.1 XM_017011295.1 XM_017011296.1 XM_017011297.1 XM_017011298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7448967 c.2089C>T p.P697S Chr.6: 31642358-31642358 on Build GRCh38
4933807 c.2152C>T p.P718S Chr.6: 31642295-31642295 on Build GRCh38
4918204 c.1998G>T p.Q666H Chr.6: 31642341-31642341 on Build GRCh38
4933806 c.2044C>T p.P682S Chr.6: 31642295-31642295 on Build GRCh38
1077622 c.2034C>T p.G678G Chr.6: 31642305-31642305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766613142 Chr.6: 31642383-31642383 on Build GRCh38
rs200073459 Chr.6: 31642332-31642332 on Build GRCh38
rs770546772 Chr.6: 31642312-31642312 on Build GRCh38
rs148186069 Chr.6: 31642305-31642305 on Build GRCh38
rs753986543 Chr.6: 31642386-31642386 on Build GRCh38
rs3034254 Chr.6: 31642416-31642418 on Build GRCh38
rs747605981 Chr.6: 31642328-31642328 on Build GRCh38
rs961050537 Chr.6: 31642264-31642264 on Build GRCh38
rs752719899 Chr.6: 31642278-31642278 on Build GRCh38
rs767765376 Chr.6: 31642349-31642349 on Build GRCh38
rs557448963 Chr.6: 31642353-31642353 on Build GRCh38
rs745374375 Chr.6: 31642320-31642320 on Build GRCh38
rs758242824 Chr.6: 31642282-31642282 on Build GRCh38
rs762089651 Chr.6: 31642345-31642345 on Build GRCh38
rs949007956 Chr.6: 31642412-31642412 on Build GRCh38
rs372412369 Chr.6: 31642419-31642422 on Build GRCh38
rs774659500 Chr.6: 31642341-31642341 on Build GRCh38
rs112889368 Chr.6: 31642416-31642416 on Build GRCh38
rs534596230 Chr.6: 31642263-31642263 on Build GRCh38
rs1040906155 Chr.6: 31642421-31642421 on Build GRCh38
rs775947992 Chr.6: 31642331-31642331 on Build GRCh38
rs910789449 Chr.6: 31642344-31642344 on Build GRCh38
rs760775657 Chr.6: 31642382-31642382 on Build GRCh38
rs750677124 Chr.6: 31642374-31642374 on Build GRCh38
rs36173461 Chr.6: 31642413-31642416 on Build GRCh38
rs986776721 Chr.6: 31642277-31642277 on Build GRCh38
rs756204588 Chr.6: 31642271-31642271 on Build GRCh38
rs746828364 Chr.6: 31642307-31642307 on Build GRCh38
rs780725341 Chr.6: 31642315-31642315 on Build GRCh38
rs778903271 Chr.6: 31642273-31642273 on Build GRCh38
rs542084968 Chr.6: 31642335-31642335 on Build GRCh38
rs920101732 Chr.6: 31642324-31642324 on Build GRCh38

View Full Product Details