Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 138132558-138132831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM135B
Entrez Gene ID
51059
Gene Name
family with sequence similarity 135 member B
Gene Aliases
C8ORFK32
UniGene
Hs.126024
Gene Chromosome Location
Chr.8: 138130023-138498390 on Build GRCh38
Transcripts
NM_015912.3 XM_011517056.2 XM_011517058.2 XM_011517059.1 XM_011517060.2 XM_011517061.2 XM_011517062.1 XM_011517063.2 XM_011517064.2 XM_011517065.1 XM_011517068.2 XM_011517069.2 XM_011517072.2 XM_011517073.2 XM_011517074.1 XM_017013470.1 XM_017013471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5408877 c.4048C>T p.L1350L Chr.8: 138132766-138132766 on Build GRCh38
5408880 c.4016-1G>A p.? Chr.8: 138132799-138132799 on Build GRCh38
6516642 c.4198G>T p.A1400S Chr.8: 138132616-138132616 on Build GRCh38
3318794 c.4135G>T p.G1379C Chr.8: 138132679-138132679 on Build GRCh38
3318785 c.4206C>G p.L1402L Chr.8: 138132608-138132608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545712386 Chr.8: 138132695-138132695 on Build GRCh38
rs367883703 Chr.8: 138132710-138132710 on Build GRCh38
rs371168199 Chr.8: 138132753-138132753 on Build GRCh38
rs963566320 Chr.8: 138132733-138132733 on Build GRCh38
rs193010937 Chr.8: 138132788-138132788 on Build GRCh38
rs765497247 Chr.8: 138132719-138132720 on Build GRCh38
rs750596567 Chr.8: 138132676-138132676 on Build GRCh38
rs375563526 Chr.8: 138132662-138132662 on Build GRCh38
rs765467195 Chr.8: 138132608-138132608 on Build GRCh38
rs572655031 Chr.8: 138132661-138132661 on Build GRCh38
rs997576188 Chr.8: 138132675-138132675 on Build GRCh38
rs776299514 Chr.8: 138132720-138132720 on Build GRCh38
rs374141412 Chr.8: 138132763-138132763 on Build GRCh38
rs770522492 Chr.8: 138132719-138132719 on Build GRCh38
rs1057256014 Chr.8: 138132688-138132688 on Build GRCh38
rs775932087 Chr.8: 138132614-138132614 on Build GRCh38
rs371041013 Chr.8: 138132680-138132680 on Build GRCh38
rs745456353 Chr.8: 138132724-138132724 on Build GRCh38
rs201744533 Chr.8: 138132743-138132743 on Build GRCh38
rs1000901968 Chr.8: 138132622-138132622 on Build GRCh38
rs368501599 Chr.8: 138132778-138132778 on Build GRCh38
rs764651487 Chr.8: 138132658-138132658 on Build GRCh38
rs2280848 Chr.8: 138132671-138132671 on Build GRCh38
rs199557562 Chr.8: 138132625-138132625 on Build GRCh38
rs746980035 Chr.8: 138132718-138132718 on Build GRCh38
rs758184042 Chr.8: 138132698-138132698 on Build GRCh38
rs374966721 Chr.8: 138132694-138132694 on Build GRCh38
rs759290697 Chr.8: 138132606-138132606 on Build GRCh38
rs758974242 Chr.8: 138132727-138132727 on Build GRCh38
rs764408584 Chr.8: 138132757-138132757 on Build GRCh38
rs769486878 Chr.8: 138132742-138132742 on Build GRCh38
rs774800983 Chr.8: 138132759-138132759 on Build GRCh38
rs776388580 Chr.8: 138132599-138132599 on Build GRCh38

View Full Product Details