Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 24698855-24699128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DFNA5
Entrez Gene ID
1687
Gene Name
DFNA5, deafness associated tumor suppressor
Gene Aliases
ICERE-1
UniGene
Hs.520708
Gene Chromosome Location
Chr.7: 24698355-24758020 on Build GRCh38
Transcripts
NM_001127453.1 NM_001127454.1 NM_004403.2 XM_017011802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
364057 c.1443T>C p.L481L Chr.7: 24699074-24699074 on Build GRCh38
5611917 c.1433C>T p.P478L Chr.7: 24699084-24699084 on Build GRCh38
270918 c.1406T>A p.V469D Chr.7: 24699111-24699111 on Build GRCh38
342012 c.1467T>A p.C489* Chr.7: 24699050-24699050 on Build GRCh38
3026456 c.1389G>A p.K463K Chr.7: 24699128-24699128 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs565886081 Chr.7: 24699060-24699060 on Build GRCh38
rs764371633 Chr.7: 24698953-24698953 on Build GRCh38
rs17274530 Chr.7: 24698996-24698996 on Build GRCh38
rs889168369 Chr.7: 24698908-24698908 on Build GRCh38
rs753419040 Chr.7: 24698980-24698980 on Build GRCh38
rs985114463 Chr.7: 24698911-24698911 on Build GRCh38
rs1016635845 Chr.7: 24698938-24698938 on Build GRCh38
rs774476375 Chr.7: 24699021-24699021 on Build GRCh38
rs10266655 Chr.7: 24698939-24698939 on Build GRCh38
rs781694856 Chr.7: 24699085-24699085 on Build GRCh38
rs757639010 Chr.7: 24699083-24699083 on Build GRCh38
rs946788426 Chr.7: 24699051-24699051 on Build GRCh38
rs565492253 Chr.7: 24698895-24698895 on Build GRCh38
rs760608631 Chr.7: 24699028-24699028 on Build GRCh38
rs953666845 Chr.7: 24698893-24698893 on Build GRCh38
rs929465909 Chr.7: 24698958-24698958 on Build GRCh38
rs727504956 Chr.7: 24699101-24699101 on Build GRCh38
rs752354361 Chr.7: 24699074-24699074 on Build GRCh38
rs370375336 Chr.7: 24699007-24699007 on Build GRCh38
rs916089635 Chr.7: 24699011-24699011 on Build GRCh38
rs763318789 Chr.7: 24698882-24698882 on Build GRCh38
rs199887441 Chr.7: 24699002-24699002 on Build GRCh38
rs770550890 Chr.7: 24699035-24699036 on Build GRCh38
rs375452655 Chr.7: 24698900-24698900 on Build GRCh38
rs559632895 Chr.7: 24699030-24699030 on Build GRCh38
rs746321505 Chr.7: 24699100-24699100 on Build GRCh38
rs767273037 Chr.7: 24699027-24699027 on Build GRCh38
rs761645664 Chr.7: 24699024-24699024 on Build GRCh38
rs781717512 Chr.7: 24699000-24699000 on Build GRCh38
rs529755996 Chr.7: 24698986-24698986 on Build GRCh38
rs142378204 Chr.7: 24698994-24698994 on Build GRCh38
rs764925434 Chr.7: 24699071-24699071 on Build GRCh38
rs541774350 Chr.7: 24698945-24698945 on Build GRCh38
rs776261657 Chr.7: 24699049-24699050 on Build GRCh38
rs148775918 Chr.7: 24698883-24698883 on Build GRCh38
rs115865539 Chr.7: 24698954-24698954 on Build GRCh38
rs753365638 Chr.7: 24699063-24699063 on Build GRCh38
rs868033817 Chr.7: 24698988-24698988 on Build GRCh38
rs371973328 Chr.7: 24699009-24699009 on Build GRCh38
rs549799959 Chr.7: 24698990-24698990 on Build GRCh38
rs768953528 Chr.7: 24699020-24699020 on Build GRCh38
rs759950764 Chr.7: 24699071-24699072 on Build GRCh38
rs147841962 Chr.7: 24699069-24699069 on Build GRCh38
rs767560006 Chr.7: 24699003-24699003 on Build GRCh38
rs749474334 Chr.7: 24699013-24699013 on Build GRCh38

View Full Product Details