Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 72022159-72022432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHODH
Entrez Gene ID
1723
Gene Name
dihydroorotate dehydrogenase (quinone)
Gene Aliases
DHOdehase, POADS, URA1
UniGene
Hs.654427
Gene Chromosome Location
Chr.16: 72008744-72027659 on Build GRCh38
Transcripts
NM_001361.4 XM_005255827.3 XM_005255829.3 XM_017022990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7261804 c.706-7G>A p.? Chr.16: 72022355-72022355 on Build GRCh38
4638963 c.737T>C p.V246A Chr.16: 72022393-72022393 on Build GRCh38
6437819 c.724G>A p.G242S Chr.16: 72022380-72022380 on Build GRCh38
1379668 c.730C>T p.R244W Chr.16: 72022386-72022386 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755992174 Chr.16: 72022326-72022326 on Build GRCh38
rs1046335182 Chr.16: 72022405-72022405 on Build GRCh38
rs112530508 Chr.16: 72022327-72022327 on Build GRCh38
rs1029017944 Chr.16: 72022264-72022264 on Build GRCh38
rs186794858 Chr.16: 72022265-72022265 on Build GRCh38
rs112810961 Chr.16: 72022199-72022199 on Build GRCh38
rs777676819 Chr.16: 72022337-72022337 on Build GRCh38
rs1050567455 Chr.16: 72022183-72022183 on Build GRCh38
rs547298424 Chr.16: 72022354-72022354 on Build GRCh38
rs756693219 Chr.16: 72022350-72022351 on Build GRCh38
rs1016401419 Chr.16: 72022301-72022301 on Build GRCh38
rs267606768 Chr.16: 72022386-72022386 on Build GRCh38
rs560960026 Chr.16: 72022294-72022294 on Build GRCh38
rs370663370 Chr.16: 72022396-72022396 on Build GRCh38
rs765676703 Chr.16: 72022198-72022198 on Build GRCh38
rs769159393 Chr.16: 72022348-72022348 on Build GRCh38
rs563765148 Chr.16: 72022246-72022246 on Build GRCh38
rs1002988520 Chr.16: 72022257-72022257 on Build GRCh38
rs958941175 Chr.16: 72022276-72022276 on Build GRCh38
rs936480520 Chr.16: 72022187-72022187 on Build GRCh38
rs375651151 Chr.16: 72022328-72022328 on Build GRCh38
rs373893426 Chr.16: 72022409-72022409 on Build GRCh38
rs767012109 Chr.16: 72022387-72022387 on Build GRCh38
rs749317101 Chr.16: 72022393-72022393 on Build GRCh38
rs962726364 Chr.16: 72022225-72022225 on Build GRCh38
rs766975132 Chr.16: 72022277-72022277 on Build GRCh38
rs565333133 Chr.16: 72022403-72022403 on Build GRCh38
rs545165838 Chr.16: 72022186-72022186 on Build GRCh38
rs910289148 Chr.16: 72022348-72022349 on Build GRCh38
rs753379984 Chr.16: 72022340-72022341 on Build GRCh38
rs370985761 Chr.16: 72022320-72022320 on Build GRCh38
rs1034828192 Chr.16: 72022209-72022209 on Build GRCh38
rs756968439 Chr.16: 72022412-72022412 on Build GRCh38
rs972267049 Chr.16: 72022339-72022339 on Build GRCh38
rs752212492 Chr.16: 72022286-72022286 on Build GRCh38
rs377593977 Chr.16: 72022347-72022347 on Build GRCh38
rs750085533 Chr.16: 72022402-72022402 on Build GRCh38
rs1003922303 Chr.16: 72022214-72022214 on Build GRCh38
rs954363467 Chr.16: 72022340-72022340 on Build GRCh38
rs955556989 Chr.16: 72022268-72022268 on Build GRCh38
rs939289356 Chr.16: 72022400-72022400 on Build GRCh38
rs751151568 Chr.16: 72022201-72022201 on Build GRCh38
rs761371223 Chr.16: 72022355-72022355 on Build GRCh38
rs1016566818 Chr.16: 72022224-72022224 on Build GRCh38
rs753671419 Chr.16: 72022411-72022411 on Build GRCh38
rs754648499 Chr.16: 72022213-72022213 on Build GRCh38
rs138788258 Chr.16: 72022293-72022293 on Build GRCh38
rs558763864 Chr.16: 72022227-72022227 on Build GRCh38

View Full Product Details