Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120626926-120627151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1GALT1C1
Entrez Gene ID
29071
Gene Name
C1GALT1 specific chaperone 1
Gene Aliases
C1GALT2, C38H2-L1, COSMC, HSPC067, MST143, TNPS
UniGene
Hs.643920
Gene Chromosome Location
Chr.X: 120625674-120630150 on Build GRCh38
Transcripts
NM_001011551.2 NM_152692.4

SNP Detail

SNP ID Chromosome Location
rs144583240 Chr.X: 120627031-120627031 on Build GRCh38
rs374944944 Chr.X: 120627081-120627081 on Build GRCh38
rs745319694 Chr.X: 120627026-120627026 on Build GRCh38
rs749791934 Chr.X: 120627032-120627032 on Build GRCh38
rs369355943 Chr.X: 120627102-120627102 on Build GRCh38
rs202093233 Chr.X: 120626959-120626959 on Build GRCh38
rs371463304 Chr.X: 120627015-120627015 on Build GRCh38
rs755578000 Chr.X: 120626988-120626988 on Build GRCh38
rs773569689 Chr.X: 120627072-120627072 on Build GRCh38
rs372933859 Chr.X: 120627110-120627110 on Build GRCh38
rs755636243 Chr.X: 120627028-120627028 on Build GRCh38
rs772296238 Chr.X: 120627071-120627071 on Build GRCh38
rs1051207343 Chr.X: 120627121-120627121 on Build GRCh38
rs751364104 Chr.X: 120626994-120626994 on Build GRCh38
rs762929191 Chr.X: 120627109-120627109 on Build GRCh38
rs137853598 Chr.X: 120626965-120626965 on Build GRCh38
rs890001394 Chr.X: 120627129-120627129 on Build GRCh38
rs765196561 Chr.X: 120626969-120626969 on Build GRCh38
rs756889624 Chr.X: 120627012-120627012 on Build GRCh38
rs202177482 Chr.X: 120626957-120626957 on Build GRCh38
rs773375422 Chr.X: 120626964-120626964 on Build GRCh38
rs748483730 Chr.X: 120627054-120627054 on Build GRCh38
rs754158304 Chr.X: 120627062-120627062 on Build GRCh38
rs146307271 Chr.X: 120627097-120627097 on Build GRCh38
rs767681789 Chr.X: 120626962-120626962 on Build GRCh38
rs768653322 Chr.X: 120627049-120627049 on Build GRCh38
rs751333183 Chr.X: 120627118-120627118 on Build GRCh38
rs200602772 Chr.X: 120627009-120627013 on Build GRCh38
rs774754459 Chr.X: 120627053-120627053 on Build GRCh38
rs763941184 Chr.X: 120626989-120626989 on Build GRCh38

View Full Product Details