Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 101178572-101178845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHSY1
Entrez Gene ID
22856
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
CHSY, CSS1, ChSy-1, TPBS
UniGene
Hs.110488
Gene Chromosome Location
Chr.15: 101175723-101251932 on Build GRCh38
Transcripts
NM_014918.4 XM_006720435.3 XM_011521364.2 XM_017022011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4418915 c.1052A>G p.K351R Chr.15: 101178745-101178745 on Build GRCh38
4416978 c.1075C>T p.P359S Chr.15: 101178722-101178722 on Build GRCh38
5383035 c.1070G>A p.G357E Chr.15: 101178727-101178727 on Build GRCh38
4053520 c.995T>C p.I332T Chr.15: 101178802-101178802 on Build GRCh38
1375713 c.976G>A p.E326K Chr.15: 101178821-101178821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149429037 Chr.15: 101178645-101178645 on Build GRCh38
rs74752435 Chr.15: 101178745-101178745 on Build GRCh38
rs752480217 Chr.15: 101178766-101178766 on Build GRCh38
rs762476576 Chr.15: 101178717-101178717 on Build GRCh38
rs142783588 Chr.15: 101178822-101178822 on Build GRCh38
rs760264432 Chr.15: 101178739-101178739 on Build GRCh38
rs143705038 Chr.15: 101178814-101178814 on Build GRCh38
rs147287474 Chr.15: 101178621-101178621 on Build GRCh38
rs139894017 Chr.15: 101178630-101178630 on Build GRCh38
rs768914522 Chr.15: 101178809-101178809 on Build GRCh38
rs768947688 Chr.15: 101178712-101178712 on Build GRCh38
rs1024887904 Chr.15: 101178779-101178779 on Build GRCh38
rs573681382 Chr.15: 101178599-101178599 on Build GRCh38
rs774636195 Chr.15: 101178811-101178811 on Build GRCh38
rs3743193 Chr.15: 101178722-101178722 on Build GRCh38
rs777695123 Chr.15: 101178789-101178789 on Build GRCh38
rs1036679938 Chr.15: 101178706-101178706 on Build GRCh38
rs780157408 Chr.15: 101178799-101178799 on Build GRCh38
rs749945441 Chr.15: 101178622-101178622 on Build GRCh38
rs781308181 Chr.15: 101178672-101178672 on Build GRCh38
rs771996972 Chr.15: 101178659-101178659 on Build GRCh38
rs888511195 Chr.15: 101178705-101178705 on Build GRCh38
rs143549949 Chr.15: 101178616-101178616 on Build GRCh38
rs754629376 Chr.15: 101178752-101178752 on Build GRCh38
rs750900475 Chr.15: 101178738-101178738 on Build GRCh38
rs769801559 Chr.15: 101178688-101178688 on Build GRCh38
rs762386598 Chr.15: 101178619-101178619 on Build GRCh38
rs745964416 Chr.15: 101178684-101178684 on Build GRCh38
rs763234060 Chr.15: 101178615-101178615 on Build GRCh38
rs144853818 Chr.15: 101178657-101178657 on Build GRCh38
rs967033322 Chr.15: 101178782-101178782 on Build GRCh38
rs778640801 Chr.15: 101178765-101178765 on Build GRCh38
rs778749871 Chr.15: 101178651-101178651 on Build GRCh38
rs775725501 Chr.15: 101178604-101178604 on Build GRCh38
rs753484761 Chr.15: 101178749-101178749 on Build GRCh38
rs773884567 Chr.15: 101178821-101178821 on Build GRCh38
rs140051393 Chr.15: 101178795-101178795 on Build GRCh38
rs552343683 Chr.15: 101178715-101178715 on Build GRCh38
rs775607768 Chr.15: 101178694-101178694 on Build GRCh38
rs779863564 Chr.15: 101178639-101178639 on Build GRCh38
rs146065837 Chr.15: 101178788-101178788 on Build GRCh38
rs753461001 Chr.15: 101178640-101178640 on Build GRCh38
rs752027638 Chr.15: 101178617-101178617 on Build GRCh38
rs763183997 Chr.15: 101178710-101178710 on Build GRCh38
rs1001593723 Chr.15: 101178719-101178719 on Build GRCh38
rs769991148 Chr.15: 101178600-101178600 on Build GRCh38
rs1027407186 Chr.15: 101178695-101178695 on Build GRCh38
rs772497432 Chr.15: 101178815-101178815 on Build GRCh38
rs546352507 Chr.15: 101178774-101178774 on Build GRCh38
rs776665009 Chr.15: 101178591-101178591 on Build GRCh38
rs746911502 Chr.15: 101178792-101178792 on Build GRCh38
rs944258648 Chr.15: 101178629-101178629 on Build GRCh38
rs749508854 Chr.15: 101178808-101178808 on Build GRCh38
rs765848875 Chr.15: 101178824-101178824 on Build GRCh38

View Full Product Details