Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162189597-162189848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAP
Entrez Gene ID
2191
Gene Name
fibroblast activation protein alpha
Gene Aliases
DPPIV, FAPA, SIMP
UniGene
Hs.654370
Gene Chromosome Location
Chr.2: 162170684-162243535 on Build GRCh38
Transcripts
NM_001291807.1 NM_004460.3 XM_011510796.2 XM_011510797.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5587783 c.1451-1G>T p.? Chr.2: 162189755-162189755 on Build GRCh38
4527496 c.1468G>A p.E490K Chr.2: 162189737-162189737 on Build GRCh38
3311288 c.1511C>T p.P504L Chr.2: 162189694-162189694 on Build GRCh38
7236598 c.1467A>C p.E489D Chr.2: 162189738-162189738 on Build GRCh38
6388264 c.1543G>T p.E515* Chr.2: 162189662-162189662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769383774 Chr.2: 162189790-162189790 on Build GRCh38
rs749363185 Chr.2: 162189625-162189625 on Build GRCh38
rs760366042 Chr.2: 162189744-162189744 on Build GRCh38
rs1056272648 Chr.2: 162189763-162189763 on Build GRCh38
rs748620563 Chr.2: 162189785-162189785 on Build GRCh38
rs764847122 Chr.2: 162189694-162189694 on Build GRCh38
rs878957270 Chr.2: 162189739-162189739 on Build GRCh38
rs192963947 Chr.2: 162189745-162189745 on Build GRCh38
rs781678111 Chr.2: 162189783-162189783 on Build GRCh38
rs950074353 Chr.2: 162189817-162189817 on Build GRCh38
rs775864096 Chr.2: 162189657-162189657 on Build GRCh38
rs761385741 Chr.2: 162189690-162189690 on Build GRCh38
rs547572967 Chr.2: 162189700-162189700 on Build GRCh38
rs772364783 Chr.2: 162189656-162189656 on Build GRCh38
rs754775708 Chr.2: 162189712-162189712 on Build GRCh38
rs956870440 Chr.2: 162189638-162189638 on Build GRCh38
rs530148388 Chr.2: 162189798-162189798 on Build GRCh38
rs780190619 Chr.2: 162189752-162189752 on Build GRCh38
rs147933561 Chr.2: 162189678-162189678 on Build GRCh38
rs772875835 Chr.2: 162189793-162189793 on Build GRCh38
rs778609076 Chr.2: 162189804-162189805 on Build GRCh38
rs913528513 Chr.2: 162189623-162189623 on Build GRCh38
rs896329904 Chr.2: 162189791-162189791 on Build GRCh38
rs926544395 Chr.2: 162189714-162189714 on Build GRCh38
rs746050369 Chr.2: 162189732-162189732 on Build GRCh38
rs757337304 Chr.2: 162189635-162189635 on Build GRCh38
rs746093509 Chr.2: 162189653-162189653 on Build GRCh38
rs779102145 Chr.2: 162189651-162189651 on Build GRCh38
rs201724480 Chr.2: 162189722-162189722 on Build GRCh38
rs756859700 Chr.2: 162189757-162189758 on Build GRCh38
rs778977625 Chr.2: 162189724-162189724 on Build GRCh38
rs751334991 Chr.2: 162189702-162189702 on Build GRCh38
rs753382509 Chr.2: 162189622-162189625 on Build GRCh38
rs770682153 Chr.2: 162189800-162189800 on Build GRCh38
rs752642706 Chr.2: 162189715-162189715 on Build GRCh38
rs768970439 Chr.2: 162189764-162189764 on Build GRCh38
rs762712988 Chr.2: 162189701-162189701 on Build GRCh38
rs747387832 Chr.2: 162189753-162189753 on Build GRCh38
rs939801618 Chr.2: 162189807-162189807 on Build GRCh38
rs750681495 Chr.2: 162189677-162189677 on Build GRCh38
rs765591209 Chr.2: 162189775-162189776 on Build GRCh38
rs777064536 Chr.2: 162189689-162189689 on Build GRCh38
rs745350021 Chr.2: 162189805-162189805 on Build GRCh38
rs1009282555 Chr.2: 162189641-162189641 on Build GRCh38

View Full Product Details