Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 39950488-39950735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK13
Entrez Gene ID
8621
Gene Name
cyclin dependent kinase 13
Gene Aliases
CDC2L, CDC2L5, CHED, hCDK13
UniGene
Hs.233552
Gene Chromosome Location
Chr.7: 39949501-40097134 on Build GRCh38
Transcripts
NM_003718.4 NM_031267.3 XM_011515597.2 XM_017012750.1 XM_017012751.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1200539 c.37G>A p.G13R Chr.7: 39950678-39950678 on Build GRCh38
5732194 c.71A>C p.E24A Chr.7: 39950712-39950712 on Build GRCh38
4639569 c.30G>A p.G10G Chr.7: 39950671-39950671 on Build GRCh38
6415220 c.4C>T p.P2S Chr.7: 39950645-39950645 on Build GRCh38
4614363 c.56_58delAGA p.K21delK Chr.7: 39950697-39950699 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1037332770 Chr.7: 39950524-39950524 on Build GRCh38
rs775323128 Chr.7: 39950705-39950705 on Build GRCh38
rs976683467 Chr.7: 39950612-39950612 on Build GRCh38
rs1014302436 Chr.7: 39950530-39950530 on Build GRCh38
rs866695558 Chr.7: 39950513-39950513 on Build GRCh38
rs924722563 Chr.7: 39950525-39950525 on Build GRCh38
rs890236092 Chr.7: 39950629-39950629 on Build GRCh38
rs770994713 Chr.7: 39950634-39950634 on Build GRCh38
rs954122065 Chr.7: 39950623-39950623 on Build GRCh38
rs776666620 Chr.7: 39950640-39950640 on Build GRCh38
rs977908667 Chr.7: 39950521-39950521 on Build GRCh38
rs896830745 Chr.7: 39950529-39950529 on Build GRCh38
rs1022252618 Chr.7: 39950701-39950701 on Build GRCh38
rs1049548407 Chr.7: 39950512-39950512 on Build GRCh38
rs958558494 Chr.7: 39950570-39950570 on Build GRCh38
rs1021597124 Chr.7: 39950586-39950586 on Build GRCh38
rs1017286780 Chr.7: 39950683-39950683 on Build GRCh38
rs528605064 Chr.7: 39950545-39950545 on Build GRCh38
rs941269757 Chr.7: 39950522-39950522 on Build GRCh38
rs942245018 Chr.7: 39950601-39950601 on Build GRCh38
rs966604839 Chr.7: 39950509-39950509 on Build GRCh38
rs1011178442 Chr.7: 39950584-39950584 on Build GRCh38
rs571636079 Chr.7: 39950711-39950711 on Build GRCh38
rs17496116 Chr.7: 39950535-39950535 on Build GRCh38
rs551765285 Chr.7: 39950635-39950635 on Build GRCh38
rs1033832359 Chr.7: 39950552-39950552 on Build GRCh38
rs905606401 Chr.7: 39950538-39950538 on Build GRCh38
rs1029742495 Chr.7: 39950617-39950617 on Build GRCh38
rs1011234458 Chr.7: 39950689-39950689 on Build GRCh38
rs745696751 Chr.7: 39950684-39950684 on Build GRCh38
rs915069129 Chr.7: 39950507-39950507 on Build GRCh38
rs985289890 Chr.7: 39950646-39950646 on Build GRCh38
rs966745150 Chr.7: 39950594-39950594 on Build GRCh38
rs769526109 Chr.7: 39950688-39950688 on Build GRCh38

View Full Product Details