Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 94517481-94517726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASD1
Entrez Gene ID
64921
Gene Name
CAS1 domain containing 1
Gene Aliases
C7orf12, NBLA04196
UniGene
Hs.260041
Gene Chromosome Location
Chr.7: 94509858-94633465 on Build GRCh38
Transcripts
NM_022900.4 XM_006716093.3 XM_011516495.2 XM_017012546.1 XM_017012547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3833517 c.183G>T p.E61D Chr.7: 94517609-94517609 on Build GRCh38
1093274 c.230+2T>C p.? Chr.7: 94517658-94517658 on Build GRCh38
150328 c.230+7T>G p.? Chr.7: 94517663-94517663 on Build GRCh38
3924267 c.143C>T p.S48L Chr.7: 94517569-94517569 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747996084 Chr.7: 94517618-94517618 on Build GRCh38
rs199978470 Chr.7: 94517690-94517690 on Build GRCh38
rs745931185 Chr.7: 94517632-94517632 on Build GRCh38
rs751009280 Chr.7: 94517693-94517693 on Build GRCh38
rs188425057 Chr.7: 94517553-94517553 on Build GRCh38
rs926101751 Chr.7: 94517634-94517634 on Build GRCh38
rs751315598 Chr.7: 94517579-94517579 on Build GRCh38
rs576996718 Chr.7: 94517576-94517576 on Build GRCh38
rs977025603 Chr.7: 94517536-94517536 on Build GRCh38
rs780784679 Chr.7: 94517615-94517615 on Build GRCh38
rs370877329 Chr.7: 94517692-94517692 on Build GRCh38
rs761009514 Chr.7: 94517525-94517525 on Build GRCh38
rs754231400 Chr.7: 94517538-94517538 on Build GRCh38
rs200966305 Chr.7: 94517671-94517671 on Build GRCh38
rs749883611 Chr.7: 94517569-94517569 on Build GRCh38
rs774415976 Chr.7: 94517673-94517675 on Build GRCh38
rs1056050269 Chr.7: 94517681-94517681 on Build GRCh38
rs533332630 Chr.7: 94517635-94517635 on Build GRCh38
rs754692582 Chr.7: 94517594-94517594 on Build GRCh38
rs373549100 Chr.7: 94517626-94517626 on Build GRCh38
rs777806221 Chr.7: 94517628-94517628 on Build GRCh38
rs371003135 Chr.7: 94517622-94517622 on Build GRCh38
rs2301646 Chr.7: 94517663-94517663 on Build GRCh38
rs550913016 Chr.7: 94517629-94517629 on Build GRCh38
rs758026067 Chr.7: 94517570-94517570 on Build GRCh38
rs764382308 Chr.7: 94517532-94517532 on Build GRCh38
rs199755372 Chr.7: 94517563-94517563 on Build GRCh38
rs760817168 Chr.7: 94517660-94517660 on Build GRCh38

View Full Product Details