Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33581437-33581664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DONSON
Entrez Gene ID
29980
Gene Name
downstream neighbor of SON
Gene Aliases
B17, C21orf60
UniGene
Hs.436341
Gene Chromosome Location
Chr.21: 33577551-33588708 on Build GRCh38
Transcripts
NM_017613.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1030389 c.1214G>A p.G405E Chr.21: 33581438-33581438 on Build GRCh38
1030390 c.1190C>A p.P397H Chr.21: 33581462-33581462 on Build GRCh38
5677316 c.1177A>G p.M393V Chr.21: 33581475-33581475 on Build GRCh38
1250592 c.1153C>T p.R385C Chr.21: 33581499-33581499 on Build GRCh38
1632463 c.1206G>T p.L402F Chr.21: 33581446-33581446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902666698 Chr.21: 33581633-33581633 on Build GRCh38
rs746401552 Chr.21: 33581538-33581538 on Build GRCh38
rs946816153 Chr.21: 33581622-33581622 on Build GRCh38
rs764117179 Chr.21: 33581523-33581523 on Build GRCh38
rs750390868 Chr.21: 33581498-33581498 on Build GRCh38
rs1054300649 Chr.21: 33581596-33581596 on Build GRCh38
rs758139862 Chr.21: 33581499-33581499 on Build GRCh38
rs199575906 Chr.21: 33581560-33581560 on Build GRCh38
rs751148410 Chr.21: 33581506-33581506 on Build GRCh38
rs760980116 Chr.21: 33581480-33581480 on Build GRCh38
rs200923236 Chr.21: 33581503-33581503 on Build GRCh38
rs181879927 Chr.21: 33581587-33581587 on Build GRCh38
rs754622038 Chr.21: 33581520-33581520 on Build GRCh38
rs368214290 Chr.21: 33581588-33581588 on Build GRCh38
rs554610235 Chr.21: 33581523-33581523 on Build GRCh38
rs761844229 Chr.21: 33581494-33581494 on Build GRCh38
rs765189899 Chr.21: 33581495-33581495 on Build GRCh38
rs769231728 Chr.21: 33581524-33581524 on Build GRCh38
rs878873678 Chr.21: 33581598-33581598 on Build GRCh38
rs758411809 Chr.21: 33581488-33581495 on Build GRCh38
rs142081989 Chr.21: 33581483-33581483 on Build GRCh38
rs867411388 Chr.21: 33581502-33581502 on Build GRCh38
rs779438610 Chr.21: 33581537-33581537 on Build GRCh38
rs201828095 Chr.21: 33581486-33581486 on Build GRCh38
rs780766265 Chr.21: 33581521-33581521 on Build GRCh38
rs752405565 Chr.21: 33581467-33581467 on Build GRCh38

View Full Product Details