Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62819137-62819378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL9A3
Entrez Gene ID
1299
Gene Name
collagen type IX alpha 3
Gene Aliases
DJ885L7.4.1, EDM3, IDD, MED
UniGene
Hs.716639
Gene Chromosome Location
Chr.20: 62817062-62841159 on Build GRCh38
Transcripts
NM_001853.3 XM_011528545.1 XM_017027666.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6894920 c.223G>C p.E75Q Chr.20: 62819261-62819261 on Build GRCh38
1412996 c.213G>A p.G71G Chr.20: 62819251-62819251 on Build GRCh38
1172737 c.242T>C p.L81P Chr.20: 62819280-62819280 on Build GRCh38
6267520 c.230G>T p.G77V Chr.20: 62819268-62819268 on Build GRCh38
6633866 c.225G>T p.E75D Chr.20: 62819263-62819263 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776846997 Chr.20: 62819337-62819337 on Build GRCh38
rs371586388 Chr.20: 62819330-62819330 on Build GRCh38
rs572310112 Chr.20: 62819160-62819160 on Build GRCh38
rs754198743 Chr.20: 62819233-62819233 on Build GRCh38
rs764253144 Chr.20: 62819333-62819333 on Build GRCh38
rs143307835 Chr.20: 62819247-62819247 on Build GRCh38
rs868763684 Chr.20: 62819231-62819231 on Build GRCh38
rs200099311 Chr.20: 62819332-62819332 on Build GRCh38
rs141960487 Chr.20: 62819275-62819275 on Build GRCh38
rs368252527 Chr.20: 62819274-62819274 on Build GRCh38
rs567353466 Chr.20: 62819318-62819318 on Build GRCh38
rs985263959 Chr.20: 62819222-62819222 on Build GRCh38
rs911107185 Chr.20: 62819232-62819232 on Build GRCh38
rs202016257 Chr.20: 62819354-62819355 on Build GRCh38
rs376155030 Chr.20: 62819202-62819202 on Build GRCh38
rs754174885 Chr.20: 62819294-62819294 on Build GRCh38
rs906822613 Chr.20: 62819261-62819261 on Build GRCh38
rs199653123 Chr.20: 62819283-62819283 on Build GRCh38
rs750783999 Chr.20: 62819343-62819343 on Build GRCh38
rs886038500 Chr.20: 62819313-62819314 on Build GRCh38
rs531320203 Chr.20: 62819188-62819188 on Build GRCh38
rs759699511 Chr.20: 62819279-62819279 on Build GRCh38
rs768383885 Chr.20: 62819186-62819186 on Build GRCh38
rs112298815 Chr.20: 62819336-62819336 on Build GRCh38
rs34733117 Chr.20: 62819188-62819189 on Build GRCh38
rs758517077 Chr.20: 62819270-62819278 on Build GRCh38
rs146260681 Chr.20: 62819284-62819284 on Build GRCh38
rs758893953 Chr.20: 62819314-62819314 on Build GRCh38
rs564992042 Chr.20: 62819248-62819248 on Build GRCh38
rs551957370 Chr.20: 62819269-62819269 on Build GRCh38
rs1004200823 Chr.20: 62819350-62819350 on Build GRCh38
rs756387351 Chr.20: 62819206-62819206 on Build GRCh38
rs1048482123 Chr.20: 62819319-62819319 on Build GRCh38
rs772867026 Chr.20: 62819324-62819324 on Build GRCh38
rs768298010 Chr.20: 62819256-62819256 on Build GRCh38
rs765691301 Chr.20: 62819339-62819339 on Build GRCh38
rs778066317 Chr.20: 62819210-62819210 on Build GRCh38
rs200945571 Chr.20: 62819329-62819330 on Build GRCh38
rs372141089 Chr.20: 62819189-62819189 on Build GRCh38
rs757752596 Chr.20: 62819234-62819234 on Build GRCh38
rs748201881 Chr.20: 62819185-62819185 on Build GRCh38
rs560905601 Chr.20: 62819191-62819191 on Build GRCh38
rs769572529 Chr.20: 62819322-62819322 on Build GRCh38
rs779295260 Chr.20: 62819307-62819307 on Build GRCh38
rs373876810 Chr.20: 62819356-62819356 on Build GRCh38
rs929613477 Chr.20: 62819165-62819165 on Build GRCh38
rs369434341 Chr.20: 62819338-62819338 on Build GRCh38
rs776474577 Chr.20: 62819187-62819187 on Build GRCh38
rs537873114 Chr.20: 62819327-62819327 on Build GRCh38
rs148233256 Chr.20: 62819190-62819190 on Build GRCh38
rs538712069 Chr.20: 62819344-62819344 on Build GRCh38
rs779607039 Chr.20: 62819315-62819315 on Build GRCh38
rs749302170 Chr.20: 62819325-62819325 on Build GRCh38
rs929991533 Chr.20: 62819313-62819313 on Build GRCh38
rs549398509 Chr.20: 62819316-62819316 on Build GRCh38
rs377557569 Chr.20: 62819308-62819309 on Build GRCh38
rs368003009 Chr.20: 62819170-62819170 on Build GRCh38
rs1052726055 Chr.20: 62819355-62819355 on Build GRCh38
rs760469617 Chr.20: 62819175-62819175 on Build GRCh38
rs532209302 Chr.20: 62819268-62819268 on Build GRCh38
rs780852369 Chr.20: 62819317-62819317 on Build GRCh38
rs988501579 Chr.20: 62819162-62819162 on Build GRCh38
rs1054305077 Chr.20: 62819311-62819311 on Build GRCh38
rs1010921416 Chr.20: 62819282-62819282 on Build GRCh38
rs764354782 Chr.20: 62819288-62819288 on Build GRCh38
rs141737082 Chr.20: 62819301-62819301 on Build GRCh38
rs745402497 Chr.20: 62819238-62819238 on Build GRCh38

View Full Product Details