Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133154037-133154298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBGT1
Entrez Gene ID
26301
Gene Name
globoside alpha-1,3-N-acetylgalactosaminyltransferase 1
Gene Aliases
A3GALNT, FS, UNQ2513
UniGene
Hs.495419
Gene Chromosome Location
Chr.9: 133152948-133163945 on Build GRCh38
Transcripts
NM_001282629.1 NM_001282632.1 NM_001288572.1 NM_001288573.1 NM_021996.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
161221 c.447C>T p.D149D Chr.9: 133154174-133154174 on Build GRCh38
3847953 c.536C>T p.T179I Chr.9: 133154085-133154085 on Build GRCh38
6213739 c.545G>A p.R182H Chr.9: 133154076-133154076 on Build GRCh38
6319826 c.568C>A p.H190N Chr.9: 133154053-133154053 on Build GRCh38
6669765 c.396C>T p.A132A Chr.9: 133154225-133154225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149153497 Chr.9: 133154074-133154074 on Build GRCh38
rs761821545 Chr.9: 133154114-133154114 on Build GRCh38
rs776225870 Chr.9: 133154188-133154188 on Build GRCh38
rs1042346887 Chr.9: 133154152-133154152 on Build GRCh38
rs373531092 Chr.9: 133154150-133154150 on Build GRCh38
rs767596461 Chr.9: 133154225-133154225 on Build GRCh38
rs376026344 Chr.9: 133154235-133154235 on Build GRCh38
rs1003658736 Chr.9: 133154156-133154156 on Build GRCh38
rs770968554 Chr.9: 133154085-133154085 on Build GRCh38
rs371591085 Chr.9: 133154107-133154107 on Build GRCh38
rs370796684 Chr.9: 133154259-133154259 on Build GRCh38
rs749842818 Chr.9: 133154063-133154063 on Build GRCh38
rs35808997 Chr.9: 133154087-133154087 on Build GRCh38
rs895356162 Chr.9: 133154195-133154195 on Build GRCh38
rs34260370 Chr.9: 133154134-133154134 on Build GRCh38
rs147110868 Chr.9: 133154148-133154148 on Build GRCh38
rs746076670 Chr.9: 133154100-133154100 on Build GRCh38
rs764506322 Chr.9: 133154205-133154205 on Build GRCh38
rs758181959 Chr.9: 133154256-133154256 on Build GRCh38
rs1017203892 Chr.9: 133154237-133154237 on Build GRCh38
rs186077979 Chr.9: 133154147-133154147 on Build GRCh38
rs545204928 Chr.9: 133154076-133154076 on Build GRCh38
rs750452166 Chr.9: 133154227-133154227 on Build GRCh38
rs747855615 Chr.9: 133154168-133154168 on Build GRCh38
rs778788525 Chr.9: 133154162-133154162 on Build GRCh38
rs750494096 Chr.9: 133154108-133154108 on Build GRCh38
rs145926254 Chr.9: 133154252-133154252 on Build GRCh38
rs147921910 Chr.9: 133154125-133154125 on Build GRCh38
rs760689441 Chr.9: 133154174-133154174 on Build GRCh38
rs376407633 Chr.9: 133154185-133154185 on Build GRCh38
rs764757622 Chr.9: 133154192-133154192 on Build GRCh38
rs567017454 Chr.9: 133154073-133154073 on Build GRCh38
rs200869632 Chr.9: 133154077-133154077 on Build GRCh38
rs746093226 Chr.9: 133154158-133154158 on Build GRCh38
rs756387643 Chr.9: 133154239-133154239 on Build GRCh38
rs955198190 Chr.9: 133154278-133154278 on Build GRCh38
rs752126496 Chr.9: 133154208-133154208 on Build GRCh38
rs759200887 Chr.9: 133154138-133154138 on Build GRCh38
rs371606263 Chr.9: 133154247-133154247 on Build GRCh38
rs756230595 Chr.9: 133154173-133154184 on Build GRCh38
rs755810451 Chr.9: 133154064-133154064 on Build GRCh38
rs1025284016 Chr.9: 133154273-133154273 on Build GRCh38
rs765650473 Chr.9: 133154139-133154139 on Build GRCh38
rs200931814 Chr.9: 133154199-133154199 on Build GRCh38
rs773068893 Chr.9: 133154171-133154171 on Build GRCh38
rs753614832 Chr.9: 133154135-133154135 on Build GRCh38
rs41297225 Chr.9: 133154133-133154133 on Build GRCh38
rs771865093 Chr.9: 133154169-133154169 on Build GRCh38
rs962912980 Chr.9: 133154257-133154257 on Build GRCh38
rs769944218 Chr.9: 133154161-133154161 on Build GRCh38
rs547647945 Chr.9: 133154155-133154155 on Build GRCh38
rs772584964 Chr.9: 133154079-133154079 on Build GRCh38
rs117609609 Chr.9: 133154224-133154224 on Build GRCh38
rs369096198 Chr.9: 133154277-133154277 on Build GRCh38
rs767056180 Chr.9: 133154062-133154062 on Build GRCh38
rs369113842 Chr.9: 133154197-133154197 on Build GRCh38

View Full Product Details