Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 41224861-41225122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNB1
Entrez Gene ID
1499
Gene Name
catenin beta 1
Gene Aliases
CTNNB, MRD19, armadillo
UniGene
Hs.476018
Gene Chromosome Location
Chr.3: 41199451-41240448 on Build GRCh38
Transcripts
NM_001098209.1 NM_001098210.1 NM_001904.3 XM_005264886.2 XM_006712983.1 XM_006712984.1 XM_006712985.1 XM_017005738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6941850 c.102_416del515 p.? Chr.3: 41224614-41225128 on Build GRCh38
6035 c.14_430del417 p.A5_Q143del Chr.3: 41224526-41225142 on Build GRCh38
446497 c.390G>A p.Q130Q Chr.3: 41225102-41225102 on Build GRCh38
6964429 c.367C>T p.Q123* Chr.3: 41225079-41225079 on Build GRCh38
6051 c.64_306del243 p.V22_T102del Chr.3: 41224576-41225018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748781625 Chr.3: 41224957-41224957 on Build GRCh38
rs763882677 Chr.3: 41225022-41225022 on Build GRCh38
rs372465029 Chr.3: 41224920-41224920 on Build GRCh38
rs549121138 Chr.3: 41224911-41224911 on Build GRCh38
rs767219370 Chr.3: 41224988-41224988 on Build GRCh38
rs752945251 Chr.3: 41225093-41225093 on Build GRCh38
rs775104326 Chr.3: 41224995-41224995 on Build GRCh38
rs759232889 Chr.3: 41224982-41224982 on Build GRCh38
rs746139399 Chr.3: 41225029-41225029 on Build GRCh38
rs772221205 Chr.3: 41224933-41224934 on Build GRCh38
rs529083869 Chr.3: 41224909-41224909 on Build GRCh38
rs747306661 Chr.3: 41224910-41224910 on Build GRCh38
rs202217100 Chr.3: 41225094-41225094 on Build GRCh38
rs909859735 Chr.3: 41224903-41224903 on Build GRCh38
rs777017789 Chr.3: 41224940-41224940 on Build GRCh38
rs751808983 Chr.3: 41225082-41225082 on Build GRCh38
rs775857557 Chr.3: 41224946-41224946 on Build GRCh38
rs770494663 Chr.3: 41224966-41224966 on Build GRCh38
rs753874922 Chr.3: 41225024-41225024 on Build GRCh38
rs745928417 Chr.3: 41224919-41224920 on Build GRCh38
rs781509994 Chr.3: 41225084-41225084 on Build GRCh38
rs748570447 Chr.3: 41224928-41224928 on Build GRCh38
rs56410740 Chr.3: 41224931-41224931 on Build GRCh38
rs773781329 Chr.3: 41224956-41224956 on Build GRCh38
rs764119976 Chr.3: 41224906-41224906 on Build GRCh38
rs758551763 Chr.3: 41225081-41225081 on Build GRCh38
rs755204384 Chr.3: 41225083-41225083 on Build GRCh38
rs773961563 Chr.3: 41224974-41224974 on Build GRCh38
rs770107882 Chr.3: 41225059-41225059 on Build GRCh38
rs758568335 Chr.3: 41224907-41224907 on Build GRCh38
rs770161910 Chr.3: 41224932-41224932 on Build GRCh38
rs760527240 Chr.3: 41225004-41225004 on Build GRCh38

View Full Product Details