Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178616815-178617058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLK4
Entrez Gene ID
57396
Gene Name
CDC like kinase 4
Gene Aliases
-
UniGene
Hs.406557
Gene Chromosome Location
Chr.5: 178602664-178627053 on Build GRCh38
Transcripts
NM_020666.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1066493 c.489C>T p.D163D Chr.5: 178616935-178616935 on Build GRCh38
737549 c.517G>C p.V173L Chr.5: 178616907-178616907 on Build GRCh38
1672165 c.529A>C p.I177L Chr.5: 178616895-178616895 on Build GRCh38
4839090 c.483C>T p.I161I Chr.5: 178616941-178616941 on Build GRCh38
737547 c.499G>A p.E167K Chr.5: 178616925-178616925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761792500 Chr.5: 178616887-178616887 on Build GRCh38
rs769637879 Chr.5: 178616998-178616998 on Build GRCh38
rs199959394 Chr.5: 178616934-178616934 on Build GRCh38
rs747227940 Chr.5: 178616974-178616974 on Build GRCh38
rs867263069 Chr.5: 178616877-178616877 on Build GRCh38
rs747861084 Chr.5: 178616990-178616990 on Build GRCh38
rs775629044 Chr.5: 178616856-178616856 on Build GRCh38
rs762488553 Chr.5: 178616895-178616895 on Build GRCh38
rs945617100 Chr.5: 178616978-178616978 on Build GRCh38
rs138880229 Chr.5: 178616917-178616917 on Build GRCh38
rs746384157 Chr.5: 178616941-178616941 on Build GRCh38
rs757669520 Chr.5: 178616935-178616935 on Build GRCh38
rs562656622 Chr.5: 178616987-178616987 on Build GRCh38
rs751244289 Chr.5: 178616909-178616909 on Build GRCh38
rs765466785 Chr.5: 178616889-178616889 on Build GRCh38
rs773316322 Chr.5: 178616893-178616893 on Build GRCh38
rs780171122 Chr.5: 178616971-178616971 on Build GRCh38
rs754626384 Chr.5: 178616911-178616911 on Build GRCh38
rs979485477 Chr.5: 178616984-178616984 on Build GRCh38
rs747285138 Chr.5: 178616865-178616865 on Build GRCh38
rs773227939 Chr.5: 178616999-178616999 on Build GRCh38
rs776936814 Chr.5: 178616989-178616989 on Build GRCh38
rs768489457 Chr.5: 178616875-178616875 on Build GRCh38
rs745906262 Chr.5: 178616846-178616846 on Build GRCh38
rs779323508 Chr.5: 178616940-178616940 on Build GRCh38
rs776335657 Chr.5: 178616883-178616883 on Build GRCh38
rs766002788 Chr.5: 178616905-178616905 on Build GRCh38
rs772141807 Chr.5: 178616849-178616849 on Build GRCh38
rs758455527 Chr.5: 178616961-178616961 on Build GRCh38

View Full Product Details