Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12689449-12689674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW9
Entrez Gene ID
84261
Gene Name
F-box and WD repeat domain containing 9
Gene Aliases
Fbw9
UniGene
Hs.515154
Gene Chromosome Location
Chr.19: 12688916-12696900 on Build GRCh38
Transcripts
NM_032301.2 XM_005260096.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5760340 c.1147-7C>A p.? Chr.19: 12689637-12689637 on Build GRCh38
5760341 c.201-7C>A p.? Chr.19: 12689637-12689637 on Build GRCh38
5493838 c.1195G>A p.V399M Chr.19: 12689582-12689582 on Build GRCh38
4788288 c.1147T>C p.S383P Chr.19: 12689630-12689630 on Build GRCh38
3528798 c.1230C>T p.T410T Chr.19: 12689547-12689547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754516876 Chr.19: 12689559-12689559 on Build GRCh38
rs765055232 Chr.19: 12689518-12689518 on Build GRCh38
rs761709954 Chr.19: 12689509-12689509 on Build GRCh38
rs530614098 Chr.19: 12689536-12689536 on Build GRCh38
rs751074062 Chr.19: 12689557-12689557 on Build GRCh38
rs764567821 Chr.19: 12689580-12689580 on Build GRCh38
rs374101267 Chr.19: 12689478-12689478 on Build GRCh38
rs758586765 Chr.19: 12689632-12689632 on Build GRCh38
rs911092820 Chr.19: 12689495-12689495 on Build GRCh38
rs950977822 Chr.19: 12689519-12689519 on Build GRCh38
rs1016403003 Chr.19: 12689592-12689592 on Build GRCh38
rs762643482 Chr.19: 12689543-12689543 on Build GRCh38
rs139718306 Chr.19: 12689547-12689547 on Build GRCh38
rs144031558 Chr.19: 12689582-12689582 on Build GRCh38
rs748147575 Chr.19: 12689473-12689473 on Build GRCh38
rs776696536 Chr.19: 12689497-12689497 on Build GRCh38
rs201221415 Chr.19: 12689492-12689492 on Build GRCh38
rs1050023376 Chr.19: 12689468-12689468 on Build GRCh38
rs993470140 Chr.19: 12689583-12689583 on Build GRCh38
rs375872729 Chr.19: 12689484-12689484 on Build GRCh38
rs566333570 Chr.19: 12689503-12689503 on Build GRCh38
rs780317159 Chr.19: 12689639-12689639 on Build GRCh38
rs747116147 Chr.19: 12689655-12689655 on Build GRCh38
rs770397351 Chr.19: 12689553-12689553 on Build GRCh38
rs746208067 Chr.19: 12689628-12689628 on Build GRCh38
rs779211088 Chr.19: 12689613-12689613 on Build GRCh38
rs757624871 Chr.19: 12689584-12689584 on Build GRCh38

View Full Product Details