Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66177053-66177282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRH
Entrez Gene ID
1392
Gene Name
corticotropin releasing hormone
Gene Aliases
CRF, CRH1
UniGene
Hs.75294
Gene Chromosome Location
Chr.8: 66176377-66186685 on Build GRCh38
Transcripts
NM_000756.3 XM_017013090.1 XM_017013091.1 XM_017013092.1 XM_017013093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4798965 c.414G>A p.R138R Chr.8: 66177064-66177064 on Build GRCh38
6635435 c.353A>T p.Q118L Chr.8: 66177125-66177125 on Build GRCh38
4826972 c.423C>G p.L141L Chr.8: 66177055-66177055 on Build GRCh38
6579779 c.396C>G p.L132L Chr.8: 66177082-66177082 on Build GRCh38
4138726 c.370C>A p.R124S Chr.8: 66177108-66177108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs865912065 Chr.8: 66177176-66177176 on Build GRCh38
rs989711323 Chr.8: 66177226-66177226 on Build GRCh38
rs780597345 Chr.8: 66177239-66177239 on Build GRCh38
rs776149245 Chr.8: 66177073-66177073 on Build GRCh38
rs79470909 Chr.8: 66177091-66177091 on Build GRCh38
rs768929387 Chr.8: 66177133-66177133 on Build GRCh38
rs112670926 Chr.8: 66177108-66177108 on Build GRCh38
rs6159 Chr.8: 66177190-66177190 on Build GRCh38
rs894269774 Chr.8: 66177112-66177112 on Build GRCh38
rs750442045 Chr.8: 66177208-66177208 on Build GRCh38
rs895640604 Chr.8: 66177236-66177236 on Build GRCh38
rs1030934653 Chr.8: 66177202-66177202 on Build GRCh38
rs964237596 Chr.8: 66177248-66177248 on Build GRCh38
rs764881373 Chr.8: 66177237-66177237 on Build GRCh38
rs766492442 Chr.8: 66177084-66177084 on Build GRCh38
rs758382209 Chr.8: 66177083-66177083 on Build GRCh38
rs754743039 Chr.8: 66177094-66177094 on Build GRCh38
rs764679955 Chr.8: 66177079-66177079 on Build GRCh38
rs769613042 Chr.8: 66177184-66177184 on Build GRCh38
rs750758925 Chr.8: 66177149-66177149 on Build GRCh38
rs766362720 Chr.8: 66177188-66177188 on Build GRCh38
rs778930754 Chr.8: 66177111-66177111 on Build GRCh38
rs543848155 Chr.8: 66177088-66177088 on Build GRCh38
rs772847264 Chr.8: 66177185-66177185 on Build GRCh38
rs993407249 Chr.8: 66177224-66177224 on Build GRCh38
rs1014469263 Chr.8: 66177238-66177238 on Build GRCh38
rs779161937 Chr.8: 66177199-66177199 on Build GRCh38
rs1026602893 Chr.8: 66177235-66177235 on Build GRCh38
rs761349409 Chr.8: 66177076-66177076 on Build GRCh38
rs759336356 Chr.8: 66177193-66177193 on Build GRCh38
rs943059776 Chr.8: 66177155-66177155 on Build GRCh38
rs749494262 Chr.8: 66177097-66177097 on Build GRCh38
rs752575694 Chr.8: 66177195-66177195 on Build GRCh38
rs746013419 Chr.8: 66177113-66177113 on Build GRCh38
rs772551722 Chr.8: 66177116-66177116 on Build GRCh38
rs970813329 Chr.8: 66177160-66177160 on Build GRCh38
rs777985451 Chr.8: 66177095-66177095 on Build GRCh38
rs367761064 Chr.8: 66177075-66177075 on Build GRCh38
rs775986889 Chr.8: 66177121-66177121 on Build GRCh38
rs762610957 Chr.8: 66177186-66177186 on Build GRCh38
rs958289292 Chr.8: 66177212-66177212 on Build GRCh38
rs757415012 Chr.8: 66177196-66177196 on Build GRCh38
rs761353791 Chr.8: 66177157-66177157 on Build GRCh38
rs763741427 Chr.8: 66177072-66177072 on Build GRCh38
rs750005001 Chr.8: 66177080-66177080 on Build GRCh38
rs761000328 Chr.8: 66177070-66177070 on Build GRCh38
rs757548677 Chr.8: 66177100-66177100 on Build GRCh38
rs1051531922 Chr.8: 66177131-66177131 on Build GRCh38

View Full Product Details