Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141641655-141641888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCHSD1
Entrez Gene ID
89848
Gene Name
FCH and double SH3 domains 1
Gene Aliases
NWK2
UniGene
Hs.591257
Gene Chromosome Location
Chr.5: 141639302-141651453 on Build GRCh38
Transcripts
NM_033449.2 XM_005268524.4 XM_006714803.3 XM_011537698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5019757 c.1982A>C p.D661A Chr.5: 141641727-141641727 on Build GRCh38
1206941 c.1990G>A p.A664T Chr.5: 141641719-141641719 on Build GRCh38
3852237 c.1978C>A p.L660M Chr.5: 141641731-141641731 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758397042 Chr.5: 141641748-141641748 on Build GRCh38
rs540865557 Chr.5: 141641844-141641844 on Build GRCh38
rs116772138 Chr.5: 141641727-141641727 on Build GRCh38
rs191033860 Chr.5: 141641847-141641847 on Build GRCh38
rs746027577 Chr.5: 141641729-141641729 on Build GRCh38
rs1004966635 Chr.5: 141641862-141641862 on Build GRCh38
rs1053515828 Chr.5: 141641771-141641771 on Build GRCh38
rs757602874 Chr.5: 141641717-141641717 on Build GRCh38
rs181718574 Chr.5: 141641706-141641706 on Build GRCh38
rs200119620 Chr.5: 141641702-141641702 on Build GRCh38
rs751126510 Chr.5: 141641701-141641701 on Build GRCh38
rs914975684 Chr.5: 141641820-141641820 on Build GRCh38
rs75142511 Chr.5: 141641739-141641739 on Build GRCh38
rs1052826450 Chr.5: 141641693-141641693 on Build GRCh38
rs756057008 Chr.5: 141641712-141641712 on Build GRCh38
rs186230756 Chr.5: 141641793-141641793 on Build GRCh38
rs906113707 Chr.5: 141641767-141641767 on Build GRCh38
rs560686372 Chr.5: 141641855-141641855 on Build GRCh38
rs544582821 Chr.5: 141641713-141641713 on Build GRCh38
rs767554840 Chr.5: 141641703-141641703 on Build GRCh38
rs780126795 Chr.5: 141641763-141641763 on Build GRCh38
rs765970890 Chr.5: 141641700-141641700 on Build GRCh38
rs564451342 Chr.5: 141641716-141641716 on Build GRCh38
rs752561214 Chr.5: 141641707-141641707 on Build GRCh38

View Full Product Details