Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92032386-92032649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHX4
Entrez Gene ID
253152
Gene Name
epoxide hydrolase 4
Gene Aliases
ABHD7, EH4, EPHXRP
UniGene
Hs.201555
Gene Chromosome Location
Chr.1: 92029976-92063536 on Build GRCh38
Transcripts
NM_173567.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7305705 c.252C>G p.H84Q Chr.1: 92032525-92032525 on Build GRCh38
1344884 c.313T>A p.F105I Chr.1: 92032586-92032586 on Build GRCh38
5705585 c.317+2T>C p.? Chr.1: 92032592-92032592 on Build GRCh38
3493341 c.266G>A p.G89E Chr.1: 92032539-92032539 on Build GRCh38
5964513 c.232G>A p.D78N Chr.1: 92032505-92032505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377421759 Chr.1: 92032549-92032549 on Build GRCh38
rs774733122 Chr.1: 92032454-92032454 on Build GRCh38
rs562388088 Chr.1: 92032489-92032489 on Build GRCh38
rs773652056 Chr.1: 92032470-92032470 on Build GRCh38
rs139290994 Chr.1: 92032563-92032563 on Build GRCh38
rs780581886 Chr.1: 92032499-92032499 on Build GRCh38
rs761240966 Chr.1: 92032472-92032472 on Build GRCh38
rs762897442 Chr.1: 92032550-92032550 on Build GRCh38
rs200101312 Chr.1: 92032462-92032462 on Build GRCh38
rs150574491 Chr.1: 92032443-92032443 on Build GRCh38
rs1037541739 Chr.1: 92032461-92032461 on Build GRCh38
rs756924693 Chr.1: 92032476-92032476 on Build GRCh38
rs753917930 Chr.1: 92032603-92032603 on Build GRCh38
rs201761910 Chr.1: 92032602-92032602 on Build GRCh38
rs780015934 Chr.1: 92032591-92032591 on Build GRCh38
rs371174975 Chr.1: 92032609-92032609 on Build GRCh38
rs764914570 Chr.1: 92032500-92032500 on Build GRCh38
rs759292966 Chr.1: 92032493-92032493 on Build GRCh38
rs370131920 Chr.1: 92032467-92032467 on Build GRCh38
rs373388269 Chr.1: 92032474-92032474 on Build GRCh38
rs751488831 Chr.1: 92032561-92032561 on Build GRCh38
rs532927913 Chr.1: 92032512-92032512 on Build GRCh38
rs1040026414 Chr.1: 92032441-92032441 on Build GRCh38
rs907965166 Chr.1: 92032430-92032430 on Build GRCh38
rs763974214 Chr.1: 92032560-92032560 on Build GRCh38
rs756305875 Chr.1: 92032543-92032543 on Build GRCh38
rs866268770 Chr.1: 92032511-92032511 on Build GRCh38

View Full Product Details