Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36222736-36222961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNE
Entrez Gene ID
10020
Gene Name
glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase
Gene Aliases
DMRV, GLCNE, IBM2, NM, Uae1
UniGene
Hs.5920
Gene Chromosome Location
Chr.9: 36214441-36277056 on Build GRCh38
Transcripts
NM_001128227.2 NM_001190383.1 NM_001190384.1 NM_001190388.1 NM_005476.5 XM_005251334.4 XM_017014167.1 XM_017014168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6242212 c.1572C>A p.G524G Chr.9: 36222823-36222823 on Build GRCh38
1208417 c.1590C>A p.D530E Chr.9: 36222913-36222913 on Build GRCh38
1155123 c.1543T>G p.C515G Chr.9: 36222852-36222852 on Build GRCh38
5445455 c.1516G>T p.D506Y Chr.9: 36222894-36222894 on Build GRCh38
1108897 c.1558T>G p.C520G Chr.9: 36222852-36222852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753313170 Chr.9: 36222816-36222816 on Build GRCh38
rs371648692 Chr.9: 36222902-36222902 on Build GRCh38
rs779893695 Chr.9: 36222880-36222880 on Build GRCh38
rs751302642 Chr.9: 36222884-36222884 on Build GRCh38
rs764698870 Chr.9: 36222839-36222839 on Build GRCh38
rs879009276 Chr.9: 36222760-36222760 on Build GRCh38
rs769940520 Chr.9: 36222766-36222766 on Build GRCh38
rs1057516798 Chr.9: 36222887-36222887 on Build GRCh38
rs1028353184 Chr.9: 36222769-36222769 on Build GRCh38
rs773608353 Chr.9: 36222770-36222770 on Build GRCh38
rs141172610 Chr.9: 36222901-36222901 on Build GRCh38
rs1057516657 Chr.9: 36222866-36222867 on Build GRCh38
rs201808007 Chr.9: 36222812-36222812 on Build GRCh38
rs986773986 Chr.9: 36222827-36222827 on Build GRCh38
rs924069649 Chr.9: 36222804-36222804 on Build GRCh38
rs912831036 Chr.9: 36222852-36222852 on Build GRCh38
rs758033732 Chr.9: 36222865-36222865 on Build GRCh38
rs369119154 Chr.9: 36222788-36222788 on Build GRCh38
rs966918577 Chr.9: 36222879-36222879 on Build GRCh38
rs112824871 Chr.9: 36222838-36222838 on Build GRCh38
rs748005215 Chr.9: 36222900-36222900 on Build GRCh38
rs749584893 Chr.9: 36222931-36222931 on Build GRCh38
rs764024703 Chr.9: 36222770-36222771 on Build GRCh38
rs748208582 Chr.9: 36222763-36222763 on Build GRCh38
rs1057516340 Chr.9: 36222794-36222801 on Build GRCh38
rs754892377 Chr.9: 36222885-36222885 on Build GRCh38
rs759962267 Chr.9: 36222810-36222810 on Build GRCh38
rs867511335 Chr.9: 36222886-36222886 on Build GRCh38
rs1024068004 Chr.9: 36222777-36222777 on Build GRCh38
rs774849278 Chr.9: 36222790-36222790 on Build GRCh38
rs781142720 Chr.9: 36222890-36222890 on Build GRCh38

View Full Product Details