Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 37990975-37991224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYPM
Entrez Gene ID
25763
Gene Name
huntingtin interacting protein M
Gene Aliases
CXorf27, HIP17
UniGene
Hs.122959
Gene Chromosome Location
Chr.X: 37990817-37991317 on Build GRCh38
Transcripts
NM_012274.1 XM_011544001.2 XM_011544002.2 XM_017029972.1 XM_017029973.1 XM_017029974.1

Gene Symbol
SYTL5
Entrez Gene ID
94122
Gene Name
synaptotagmin like 5
Gene Aliases
slp5
UniGene
Hs.662334
Gene Chromosome Location
Chr.X: 37888915-38128820 on Build GRCh38
Transcripts
NM_012274.1 XM_011544001.2 XM_011544002.2 XM_017029972.1 XM_017029973.1 XM_017029974.1

SNP Detail

SNP ID Chromosome Location
rs751625238 Chr.X: 37991074-37991074 on Build GRCh38
rs763415603 Chr.X: 37991038-37991038 on Build GRCh38
rs202191406 Chr.X: 37991087-37991087 on Build GRCh38
rs749137050 Chr.X: 37991099-37991099 on Build GRCh38
rs372723653 Chr.X: 37991039-37991039 on Build GRCh38
rs954419502 Chr.X: 37991150-37991150 on Build GRCh38
rs375020994 Chr.X: 37991142-37991142 on Build GRCh38
rs778875629 Chr.X: 37991064-37991064 on Build GRCh38
rs926824389 Chr.X: 37991095-37991095 on Build GRCh38
rs939581976 Chr.X: 37991127-37991127 on Build GRCh38
rs61737321 Chr.X: 37991132-37991132 on Build GRCh38
rs999950006 Chr.X: 37991183-37991183 on Build GRCh38
rs17146924 Chr.X: 37991010-37991010 on Build GRCh38
rs759493997 Chr.X: 37991175-37991175 on Build GRCh38
rs766813789 Chr.X: 37991156-37991156 on Build GRCh38
rs762019936 Chr.X: 37991011-37991011 on Build GRCh38
rs752065275 Chr.X: 37991168-37991168 on Build GRCh38
rs759187702 Chr.X: 37991003-37991003 on Build GRCh38
rs770988604 Chr.X: 37991110-37991110 on Build GRCh38
rs781373230 Chr.X: 37991188-37991188 on Build GRCh38
rs200480345 Chr.X: 37991131-37991131 on Build GRCh38
rs757767343 Chr.X: 37991134-37991135 on Build GRCh38
rs779056719 Chr.X: 37991116-37991116 on Build GRCh38
rs752992902 Chr.X: 37991076-37991076 on Build GRCh38
rs781203861 Chr.X: 37991075-37991075 on Build GRCh38
rs988581900 Chr.X: 37991171-37991171 on Build GRCh38
rs752769598 Chr.X: 37991189-37991189 on Build GRCh38
rs904283071 Chr.X: 37991157-37991157 on Build GRCh38
rs756328819 Chr.X: 37991203-37991203 on Build GRCh38
rs763362824 Chr.X: 37991149-37991149 on Build GRCh38
rs374389406 Chr.X: 37991130-37991130 on Build GRCh38
rs765654218 Chr.X: 37991012-37991012 on Build GRCh38
rs750833097 Chr.X: 37991013-37991013 on Build GRCh38
rs187821931 Chr.X: 37991117-37991117 on Build GRCh38
rs371386856 Chr.X: 37991094-37991094 on Build GRCh38
rs1056753546 Chr.X: 37991128-37991128 on Build GRCh38
rs768230320 Chr.X: 37991135-37991135 on Build GRCh38
rs778820341 Chr.X: 37991072-37991073 on Build GRCh38

View Full Product Details