Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 59655855-59656080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLTC
Entrez Gene ID
1213
Gene Name
clathrin heavy chain
Gene Aliases
CHC, CHC17, CLH-17, CLTCL2, Hc
UniGene
Hs.491351
Gene Chromosome Location
Chr.17: 59619689-59696956 on Build GRCh38
Transcripts
NM_001288653.1 NM_004859.3 XM_005257012.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
982090 c.917C>T p.T306I Chr.17: 59655975-59655975 on Build GRCh38
982089 c.881A>G p.Y294C Chr.17: 59655939-59655939 on Build GRCh38
1470970 c.931G>A p.A311T Chr.17: 59655989-59655989 on Build GRCh38
1242138 c.897T>C p.S299S Chr.17: 59655955-59655955 on Build GRCh38
6424369 c.808C>A p.H270N Chr.17: 59655866-59655866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187336326 Chr.17: 59655907-59655907 on Build GRCh38
rs753103346 Chr.17: 59655976-59655976 on Build GRCh38
rs752135383 Chr.17: 59655892-59655892 on Build GRCh38
rs756921292 Chr.17: 59655978-59655978 on Build GRCh38
rs535637632 Chr.17: 59656015-59656015 on Build GRCh38
rs755458497 Chr.17: 59655900-59655900 on Build GRCh38
rs778503398 Chr.17: 59656042-59656042 on Build GRCh38
rs141466492 Chr.17: 59655935-59655935 on Build GRCh38
rs745478221 Chr.17: 59656047-59656047 on Build GRCh38

View Full Product Details