Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71105917-71106174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf65
Entrez Gene ID
158830
Gene Name
chromosome X open reading frame 65
Gene Aliases
-
UniGene
Hs.443936
Gene Chromosome Location
Chr.X: 71103865-71106973 on Build GRCh38
Transcripts
NM_001025265.2 NR_033212.1 XM_005262244.3

SNP Detail

SNP ID Chromosome Location
rs963337813 Chr.X: 71106012-71106012 on Build GRCh38
rs138480889 Chr.X: 71106113-71106113 on Build GRCh38
rs757561171 Chr.X: 71106122-71106122 on Build GRCh38
rs145838049 Chr.X: 71106028-71106028 on Build GRCh38
rs771562771 Chr.X: 71106067-71106067 on Build GRCh38
rs370915407 Chr.X: 71105948-71105948 on Build GRCh38
rs145856763 Chr.X: 71106077-71106077 on Build GRCh38
rs765249440 Chr.X: 71106011-71106011 on Build GRCh38
rs757848987 Chr.X: 71106154-71106154 on Build GRCh38
rs746604332 Chr.X: 71106081-71106081 on Build GRCh38
rs1018370012 Chr.X: 71106119-71106119 on Build GRCh38
rs762962346 Chr.X: 71106106-71106106 on Build GRCh38
rs749760816 Chr.X: 71106149-71106149 on Build GRCh38
rs775542849 Chr.X: 71106102-71106102 on Build GRCh38
rs781482086 Chr.X: 71106035-71106035 on Build GRCh38
rs765302686 Chr.X: 71106091-71106091 on Build GRCh38
rs780562762 Chr.X: 71106144-71106144 on Build GRCh38
rs758470987 Chr.X: 71106021-71106021 on Build GRCh38
rs753830759 Chr.X: 71105965-71105965 on Build GRCh38
rs150353837 Chr.X: 71105971-71105971 on Build GRCh38
rs138762266 Chr.X: 71106031-71106031 on Build GRCh38
rs746222572 Chr.X: 71106046-71106046 on Build GRCh38
rs750827232 Chr.X: 71106135-71106135 on Build GRCh38
rs770644340 Chr.X: 71106085-71106085 on Build GRCh38
rs752887711 Chr.X: 71106016-71106016 on Build GRCh38
rs767725657 Chr.X: 71106130-71106130 on Build GRCh38
rs777904371 Chr.X: 71106027-71106027 on Build GRCh38
rs776625637 Chr.X: 71106087-71106087 on Build GRCh38
rs368758032 Chr.X: 71106062-71106062 on Build GRCh38
rs764170798 Chr.X: 71106112-71106112 on Build GRCh38
rs184815664 Chr.X: 71106051-71106051 on Build GRCh38
rs188108002 Chr.X: 71106075-71106075 on Build GRCh38
rs770353426 Chr.X: 71106047-71106047 on Build GRCh38
rs1022738978 Chr.X: 71105985-71105985 on Build GRCh38
rs201967682 Chr.X: 71106088-71106088 on Build GRCh38

View Full Product Details