Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78423727-78423996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH17
Entrez Gene ID
8632
Gene Name
dynein axonemal heavy chain 17
Gene Aliases
DNAHL1, DNEL2
UniGene
Hs.375975
Gene Chromosome Location
Chr.17: 78423697-78577430 on Build GRCh38
Transcripts
NM_173628.3 XM_011525416.2 XM_017025261.1 NM_024419.4 NR_110601.1 NR_110602.1 NR_111989.1 XM_011525487.2 XM_011525488.2 XM_011525489.2 XM_017025357.1 XM_017025358.1 XM_017025359.1 XM_017025360.1 XM_017025361.1 XM_017025362.1 XM_017025363.1 XM_017025364.1 XM_017025365.1 XM_017025366.1

Gene Symbol
PGS1
Entrez Gene ID
9489
Gene Name
phosphatidylglycerophosphate synthase 1
Gene Aliases
-
UniGene
Hs.654671
Gene Chromosome Location
Chr.17: 78378618-78424659 on Build GRCh38
Transcripts
NM_173628.3 XM_011525416.2 XM_017025261.1 NM_024419.4 NR_110601.1 NR_110602.1 NR_111989.1 XM_011525487.2 XM_011525488.2 XM_011525489.2 XM_017025357.1 XM_017025358.1 XM_017025359.1 XM_017025360.1 XM_017025361.1 XM_017025362.1 XM_017025363.1 XM_017025364.1 XM_017025365.1 XM_017025366.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4872346 c.13434C>T p.A4478A Chr.17: 78423930-78423930 on Build GRCh38
6389642 c.13436T>A p.V4479E Chr.17: 78423928-78423928 on Build GRCh38
127819 c.13311G>A p.L4437L Chr.17: 78423969-78423969 on Build GRCh38
1250346 c.13356G>A p.A4452A Chr.17: 78423924-78423924 on Build GRCh38
4070132 c.13426C>T p.L4476L Chr.17: 78423938-78423938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367916838 Chr.17: 78423858-78423858 on Build GRCh38
rs779330949 Chr.17: 78423972-78423972 on Build GRCh38
rs764815092 Chr.17: 78423959-78423960 on Build GRCh38
rs753468668 Chr.17: 78423882-78423882 on Build GRCh38
rs756856607 Chr.17: 78423883-78423883 on Build GRCh38
rs761355727 Chr.17: 78423892-78423893 on Build GRCh38
rs956448686 Chr.17: 78423815-78423815 on Build GRCh38
rs751856381 Chr.17: 78423886-78423886 on Build GRCh38
rs753590432 Chr.17: 78423862-78423863 on Build GRCh38
rs779212708 Chr.17: 78423764-78423764 on Build GRCh38
rs930506200 Chr.17: 78423785-78423785 on Build GRCh38
rs777727996 Chr.17: 78423963-78423963 on Build GRCh38
rs753334032 Chr.17: 78423934-78423934 on Build GRCh38
rs377016460 Chr.17: 78423924-78423924 on Build GRCh38
rs199669660 Chr.17: 78423965-78423965 on Build GRCh38
rs763633164 Chr.17: 78423879-78423879 on Build GRCh38
rs1000797316 Chr.17: 78423869-78423869 on Build GRCh38
rs570803591 Chr.17: 78423925-78423925 on Build GRCh38
rs143246806 Chr.17: 78423949-78423949 on Build GRCh38
rs903350683 Chr.17: 78423865-78423865 on Build GRCh38
rs544945593 Chr.17: 78423800-78423800 on Build GRCh38
rs780413522 Chr.17: 78423884-78423884 on Build GRCh38
rs562278569 Chr.17: 78423767-78423767 on Build GRCh38
rs774289227 Chr.17: 78423921-78423921 on Build GRCh38
rs927551330 Chr.17: 78423861-78423861 on Build GRCh38
rs749838869 Chr.17: 78423965-78423967 on Build GRCh38
rs755041875 Chr.17: 78423763-78423763 on Build GRCh38
rs764239849 Chr.17: 78423834-78423835 on Build GRCh38
rs866168240 Chr.17: 78423912-78423912 on Build GRCh38
rs116446606 Chr.17: 78423887-78423887 on Build GRCh38
rs777476607 Chr.17: 78423843-78423843 on Build GRCh38
rs746767108 Chr.17: 78423845-78423845 on Build GRCh38
rs748666814 Chr.17: 78423897-78423897 on Build GRCh38
rs769803281 Chr.17: 78423902-78423902 on Build GRCh38
rs979108910 Chr.17: 78423787-78423790 on Build GRCh38
rs757406187 Chr.17: 78423969-78423969 on Build GRCh38
rs781607423 Chr.17: 78423894-78423894 on Build GRCh38
rs761462566 Chr.17: 78423938-78423938 on Build GRCh38
rs372004613 Chr.17: 78423948-78423948 on Build GRCh38
rs953388197 Chr.17: 78423750-78423750 on Build GRCh38
rs760156744 Chr.17: 78423875-78423875 on Build GRCh38
rs746062866 Chr.17: 78423976-78423976 on Build GRCh38
rs72903323 Chr.17: 78423801-78423801 on Build GRCh38
rs531093474 Chr.17: 78423791-78423791 on Build GRCh38
rs752671887 Chr.17: 78423868-78423868 on Build GRCh38
rs951893698 Chr.17: 78423947-78423947 on Build GRCh38
rs771217788 Chr.17: 78423911-78423911 on Build GRCh38
rs542698237 Chr.17: 78423760-78423760 on Build GRCh38
rs112488587 Chr.17: 78423761-78423761 on Build GRCh38
rs937678019 Chr.17: 78423819-78423819 on Build GRCh38
rs919056185 Chr.17: 78423784-78423784 on Build GRCh38
rs376706067 Chr.17: 78423825-78423827 on Build GRCh38
rs113538058 Chr.17: 78423951-78423951 on Build GRCh38
rs971975444 Chr.17: 78423775-78423775 on Build GRCh38
rs775335572 Chr.17: 78423929-78423929 on Build GRCh38
rs566746938 Chr.17: 78423936-78423936 on Build GRCh38
rs1056034513 Chr.17: 78423825-78423825 on Build GRCh38
rs72914863 Chr.17: 78423903-78423903 on Build GRCh38
rs141424232 Chr.17: 78423930-78423930 on Build GRCh38
rs754163022 Chr.17: 78423890-78423890 on Build GRCh38
rs1029061933 Chr.17: 78423765-78423765 on Build GRCh38

View Full Product Details