Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27493296-27493567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC4
Entrez Gene ID
64838
Gene Name
fibronectin type III domain containing 4
Gene Aliases
FRCP1
UniGene
Hs.27836
Gene Chromosome Location
Chr.2: 27491883-27495303 on Build GRCh38
Transcripts
NM_022823.2 XM_005264499.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6032172 c.518T>C p.I173T Chr.2: 27493415-27493415 on Build GRCh38
4550640 c.505G>A p.E169K Chr.2: 27493428-27493428 on Build GRCh38
6578431 c.487C>A p.R163R Chr.2: 27493446-27493446 on Build GRCh38
3580649 c.478G>A p.D160N Chr.2: 27493455-27493455 on Build GRCh38
5780552 c.488G>A p.R163Q Chr.2: 27493445-27493445 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761244499 Chr.2: 27493348-27493348 on Build GRCh38
rs762636448 Chr.2: 27493390-27493390 on Build GRCh38
rs369284755 Chr.2: 27493520-27493520 on Build GRCh38
rs142318617 Chr.2: 27493426-27493426 on Build GRCh38
rs754430637 Chr.2: 27493429-27493429 on Build GRCh38
rs749951430 Chr.2: 27493385-27493385 on Build GRCh38
rs757232291 Chr.2: 27493431-27493431 on Build GRCh38
rs374605271 Chr.2: 27493399-27493399 on Build GRCh38
rs769661406 Chr.2: 27493474-27493474 on Build GRCh38
rs921833828 Chr.2: 27493544-27493544 on Build GRCh38
rs1048241681 Chr.2: 27493470-27493470 on Build GRCh38
rs936925366 Chr.2: 27493366-27493366 on Build GRCh38
rs376562311 Chr.2: 27493468-27493468 on Build GRCh38
rs888258522 Chr.2: 27493486-27493486 on Build GRCh38
rs779025669 Chr.2: 27493433-27493433 on Build GRCh38
rs761084397 Chr.2: 27493530-27493530 on Build GRCh38
rs750629990 Chr.2: 27493445-27493445 on Build GRCh38
rs750927309 Chr.2: 27493415-27493417 on Build GRCh38
rs780311545 Chr.2: 27493447-27493447 on Build GRCh38
rs766013210 Chr.2: 27493392-27493392 on Build GRCh38
rs763155022 Chr.2: 27493488-27493489 on Build GRCh38
rs574270733 Chr.2: 27493340-27493340 on Build GRCh38
rs866250383 Chr.2: 27493402-27493402 on Build GRCh38
rs147780937 Chr.2: 27493460-27493460 on Build GRCh38
rs772775958 Chr.2: 27493487-27493487 on Build GRCh38
rs765830362 Chr.2: 27493414-27493416 on Build GRCh38
rs751264704 Chr.2: 27493525-27493525 on Build GRCh38
rs200840735 Chr.2: 27493446-27493446 on Build GRCh38
rs762428790 Chr.2: 27493517-27493517 on Build GRCh38
rs764827593 Chr.2: 27493357-27493357 on Build GRCh38
rs758429725 Chr.2: 27493369-27493370 on Build GRCh38
rs768443253 Chr.2: 27493459-27493459 on Build GRCh38
rs756565086 Chr.2: 27493400-27493400 on Build GRCh38
rs768167526 Chr.2: 27493367-27493367 on Build GRCh38

View Full Product Details