Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 154234446-154234717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCHS2
Entrez Gene ID
54798
Gene Name
dachsous cadherin-related 2
Gene Aliases
CDH27, CDHJ, CDHR7, PCDH23, PCDHJ
UniGene
Hs.655664
Gene Chromosome Location
Chr.4: 154234375-154491778 on Build GRCh38
Transcripts
NM_017639.3 XM_011532045.1 XM_011532046.2 XM_011532049.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1203116 c.8683G>T p.E2895* Chr.4: 154234604-154234604 on Build GRCh38
5918381 c.8575C>T p.P2859S Chr.4: 154234712-154234712 on Build GRCh38
3428238 c.8750G>A p.*2917* Chr.4: 154234537-154234537 on Build GRCh38
6469349 c.8595G>A p.L2865L Chr.4: 154234692-154234692 on Build GRCh38
6561232 c.8737G>A p.E2913K Chr.4: 154234550-154234550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753578826 Chr.4: 154234547-154234547 on Build GRCh38
rs371685469 Chr.4: 154234541-154234541 on Build GRCh38
rs201311804 Chr.4: 154234562-154234562 on Build GRCh38
rs774351393 Chr.4: 154234624-154234624 on Build GRCh38
rs535864785 Chr.4: 154234684-154234684 on Build GRCh38
rs748486070 Chr.4: 154234592-154234592 on Build GRCh38
rs760604721 Chr.4: 154234514-154234514 on Build GRCh38
rs201471401 Chr.4: 154234637-154234637 on Build GRCh38
rs147859519 Chr.4: 154234542-154234542 on Build GRCh38
rs1049112791 Chr.4: 154234473-154234473 on Build GRCh38
rs779610498 Chr.4: 154234570-154234570 on Build GRCh38
rs750251473 Chr.4: 154234644-154234644 on Build GRCh38
rs770084440 Chr.4: 154234600-154234600 on Build GRCh38
rs139235484 Chr.4: 154234645-154234645 on Build GRCh38
rs753741384 Chr.4: 154234631-154234631 on Build GRCh38
rs371646071 Chr.4: 154234472-154234472 on Build GRCh38
rs961005107 Chr.4: 154234478-154234478 on Build GRCh38
rs751652739 Chr.4: 154234506-154234506 on Build GRCh38
rs759627763 Chr.4: 154234626-154234626 on Build GRCh38
rs138708352 Chr.4: 154234587-154234587 on Build GRCh38
rs754567911 Chr.4: 154234672-154234672 on Build GRCh38
rs773431667 Chr.4: 154234677-154234692 on Build GRCh38
rs764026194 Chr.4: 154234540-154234540 on Build GRCh38
rs763227598 Chr.4: 154234609-154234609 on Build GRCh38
rs1028748089 Chr.4: 154234632-154234632 on Build GRCh38
rs773498691 Chr.4: 154234607-154234607 on Build GRCh38
rs992369778 Chr.4: 154234539-154234539 on Build GRCh38
rs766465123 Chr.4: 154234497-154234497 on Build GRCh38
rs758044841 Chr.4: 154234568-154234568 on Build GRCh38
rs901635123 Chr.4: 154234673-154234673 on Build GRCh38
rs757072901 Chr.4: 154234553-154234553 on Build GRCh38
rs779586043 Chr.4: 154234649-154234649 on Build GRCh38
rs145223267 Chr.4: 154234690-154234690 on Build GRCh38
rs553907289 Chr.4: 154234693-154234693 on Build GRCh38
rs761784168 Chr.4: 154234633-154234633 on Build GRCh38
rs757141123 Chr.4: 154234561-154234562 on Build GRCh38
rs202067618 Chr.4: 154234564-154234564 on Build GRCh38
rs770164578 Chr.4: 154234594-154234594 on Build GRCh38
rs778610095 Chr.4: 154234561-154234561 on Build GRCh38
rs752121816 Chr.4: 154234612-154234612 on Build GRCh38
rs766748777 Chr.4: 154234557-154234557 on Build GRCh38
rs751062395 Chr.4: 154234663-154234663 on Build GRCh38
rs746521776 Chr.4: 154234579-154234579 on Build GRCh38
rs769226248 Chr.4: 154234585-154234585 on Build GRCh38
rs940329689 Chr.4: 154234498-154234498 on Build GRCh38
rs771128723 Chr.4: 154234611-154234611 on Build GRCh38
rs748552475 Chr.4: 154234582-154234583 on Build GRCh38

View Full Product Details