Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23756381-23756610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHP2
Entrez Gene ID
63928
Gene Name
calcineurin like EF-hand protein 2
Gene Aliases
-
UniGene
Hs.178589
Gene Chromosome Location
Chr.16: 23754627-23758951 on Build GRCh38
Transcripts
NM_022097.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2150465 c.412C>T p.Q138* Chr.16: 23756447-23756447 on Build GRCh38
6037831 c.380G>A p.R127H Chr.16: 23756415-23756415 on Build GRCh38
344416 c.383A>T p.D128V Chr.16: 23756418-23756418 on Build GRCh38
6430795 c.410T>C p.L137P Chr.16: 23756445-23756445 on Build GRCh38
6229378 c.414+6C>T p.? Chr.16: 23756455-23756455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35641939 Chr.16: 23756415-23756415 on Build GRCh38
rs972681236 Chr.16: 23756510-23756510 on Build GRCh38
rs781307565 Chr.16: 23756549-23756549 on Build GRCh38
rs73550289 Chr.16: 23756548-23756548 on Build GRCh38
rs750493101 Chr.16: 23756448-23756448 on Build GRCh38
rs752062040 Chr.16: 23756462-23756462 on Build GRCh38
rs769463598 Chr.16: 23756474-23756474 on Build GRCh38
rs765181306 Chr.16: 23756442-23756442 on Build GRCh38
rs369211227 Chr.16: 23756419-23756419 on Build GRCh38
rs887940758 Chr.16: 23756575-23756575 on Build GRCh38
rs866675308 Chr.16: 23756478-23756478 on Build GRCh38
rs373651783 Chr.16: 23756481-23756481 on Build GRCh38
rs776249507 Chr.16: 23756405-23756405 on Build GRCh38
rs762131048 Chr.16: 23756439-23756439 on Build GRCh38
rs771926950 Chr.16: 23756499-23756499 on Build GRCh38
rs547492443 Chr.16: 23756501-23756501 on Build GRCh38
rs573526780 Chr.16: 23756506-23756506 on Build GRCh38
rs145686424 Chr.16: 23756527-23756527 on Build GRCh38
rs370640357 Chr.16: 23756562-23756562 on Build GRCh38
rs748803602 Chr.16: 23756477-23756477 on Build GRCh38
rs79455352 Chr.16: 23756417-23756417 on Build GRCh38
rs758446254 Chr.16: 23756453-23756453 on Build GRCh38
rs371734934 Chr.16: 23756416-23756416 on Build GRCh38
rs772792474 Chr.16: 23756420-23756420 on Build GRCh38
rs780605025 Chr.16: 23756455-23756455 on Build GRCh38
rs759560832 Chr.16: 23756488-23756488 on Build GRCh38
rs747808324 Chr.16: 23756407-23756407 on Build GRCh38
rs1042303184 Chr.16: 23756491-23756491 on Build GRCh38
rs946565899 Chr.16: 23756468-23756468 on Build GRCh38
rs536022441 Chr.16: 23756511-23756511 on Build GRCh38
rs199831568 Chr.16: 23756420-23756420 on Build GRCh38
rs781537387 Chr.16: 23756463-23756463 on Build GRCh38
rs1027705120 Chr.16: 23756561-23756561 on Build GRCh38
rs769222851 Chr.16: 23756414-23756414 on Build GRCh38
rs774333830 Chr.16: 23756486-23756486 on Build GRCh38
rs767478843 Chr.16: 23756418-23756418 on Build GRCh38
rs918226459 Chr.16: 23756521-23756521 on Build GRCh38
rs1005502168 Chr.16: 23756588-23756588 on Build GRCh38
rs916327165 Chr.16: 23756569-23756569 on Build GRCh38
rs1056840379 Chr.16: 23756536-23756536 on Build GRCh38
rs1019482927 Chr.16: 23756473-23756473 on Build GRCh38

View Full Product Details