Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137743364-137743621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHMT1
Entrez Gene ID
79813
Gene Name
euchromatic histone lysine methyltransferase 1
Gene Aliases
EUHMTASE1, Eu-HMTase1, FP13812, GLP, GLP1, KMT1D, bA188C12.1
UniGene
Hs.495511
Gene Chromosome Location
Chr.9: 137618988-137836127 on Build GRCh38
Transcripts
NM_001145527.1 NM_024757.4 XM_005266105.4 XM_005266110.1 XM_006717288.2 XM_011519021.2 XM_011519022.2 XM_011519023.2 XM_011519024.2 XM_011519025.1 XM_011519026.2 XM_011519027.2 XM_011519028.2 XM_011519033.2 XM_017015134.1 XM_017015135.1 XM_017015136.1 XM_017015137.1 XM_017015138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5573326 c.824-1G>T p.? Chr.9: 137743370-137743370 on Build GRCh38
4372726 c.807T>C p.P269P Chr.9: 137743447-137743447 on Build GRCh38
3848125 c.877A>G p.M293V Chr.9: 137743424-137743424 on Build GRCh38
3848126 c.784A>G p.M262V Chr.9: 137743424-137743424 on Build GRCh38
5835685 c.755C>T p.A252V Chr.9: 137743395-137743395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779852017 Chr.9: 137743573-137743573 on Build GRCh38
rs149535210 Chr.9: 137743594-137743594 on Build GRCh38
rs752667977 Chr.9: 137743439-137743439 on Build GRCh38
rs771464978 Chr.9: 137743496-137743496 on Build GRCh38
rs906629447 Chr.9: 137743595-137743595 on Build GRCh38
rs142727972 Chr.9: 137743479-137743479 on Build GRCh38
rs797045557 Chr.9: 137743535-137743535 on Build GRCh38
rs749520873 Chr.9: 137743468-137743468 on Build GRCh38
rs116060838 Chr.9: 137743593-137743593 on Build GRCh38
rs778449895 Chr.9: 137743490-137743490 on Build GRCh38
rs780215981 Chr.9: 137743463-137743463 on Build GRCh38
rs766083787 Chr.9: 137743400-137743400 on Build GRCh38
rs547362155 Chr.9: 137743580-137743580 on Build GRCh38
rs772646625 Chr.9: 137743485-137743485 on Build GRCh38
rs958602333 Chr.9: 137743524-137743524 on Build GRCh38
rs529993515 Chr.9: 137743574-137743574 on Build GRCh38
rs755085175 Chr.9: 137743572-137743572 on Build GRCh38
rs375192869 Chr.9: 137743521-137743521 on Build GRCh38
rs908083992 Chr.9: 137743502-137743502 on Build GRCh38
rs762465139 Chr.9: 137743546-137743546 on Build GRCh38
rs748259939 Chr.9: 137743480-137743480 on Build GRCh38
rs75421798 Chr.9: 137743537-137743537 on Build GRCh38
rs751536740 Chr.9: 137743557-137743557 on Build GRCh38
rs747341699 Chr.9: 137743492-137743492 on Build GRCh38
rs776328408 Chr.9: 137743516-137743516 on Build GRCh38
rs755069826 Chr.9: 137743476-137743476 on Build GRCh38
rs758397921 Chr.9: 137743441-137743441 on Build GRCh38
rs137852714 Chr.9: 137743418-137743418 on Build GRCh38
rs763977391 Chr.9: 137743550-137743550 on Build GRCh38
rs565065320 Chr.9: 137743452-137743452 on Build GRCh38
rs751305840 Chr.9: 137743449-137743449 on Build GRCh38
rs376384316 Chr.9: 137743438-137743438 on Build GRCh38
rs774795859 Chr.9: 137743544-137743544 on Build GRCh38
rs544345305 Chr.9: 137743538-137743538 on Build GRCh38
rs759057973 Chr.9: 137743407-137743407 on Build GRCh38
rs920806584 Chr.9: 137743586-137743586 on Build GRCh38
rs138108260 Chr.9: 137743570-137743570 on Build GRCh38
rs764024176 Chr.9: 137743443-137743443 on Build GRCh38
rs761573493 Chr.9: 137743558-137743558 on Build GRCh38
rs867504844 Chr.9: 137743533-137743533 on Build GRCh38

View Full Product Details