Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11424214-11424485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM167A
Entrez Gene ID
83648
Gene Name
family with sequence similarity 167 member A
Gene Aliases
C8orf13, D8S265
UniGene
Hs.124299
Gene Chromosome Location
Chr.8: 11421464-11475904 on Build GRCh38
Transcripts
NM_053279.2 XM_005272398.4 XM_011543837.1 XM_011543838.2 XM_011543840.2 NR_026814.1

Gene Symbol
FAM167A-AS1
Entrez Gene ID
83656
Gene Name
FAM167A antisense RNA 1
Gene Aliases
C8orf12
UniGene
Hs.583896
Gene Chromosome Location
Chr.8: 11368402-11438657 on Build GRCh38
Transcripts
NM_053279.2 XM_005272398.4 XM_011543837.1 XM_011543838.2 XM_011543840.2 NR_026814.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6658841 c.625C>T p.R209W Chr.8: 11424393-11424393 on Build GRCh38
1643471 c.563C>A p.S188Y Chr.8: 11424455-11424455 on Build GRCh38
5408652 c.609G>A p.K203K Chr.8: 11424409-11424409 on Build GRCh38
3781362 c.534C>T p.A178A Chr.8: 11424484-11424484 on Build GRCh38
244371 c.601G>A p.V201M Chr.8: 11424417-11424417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989481310 Chr.8: 11424269-11424269 on Build GRCh38
rs989648123 Chr.8: 11424381-11424381 on Build GRCh38
rs747371658 Chr.8: 11424369-11424369 on Build GRCh38
rs1030721261 Chr.8: 11424343-11424343 on Build GRCh38
rs768510757 Chr.8: 11424454-11424454 on Build GRCh38
rs755737011 Chr.8: 11424399-11424399 on Build GRCh38
rs773785540 Chr.8: 11424323-11424341 on Build GRCh38
rs963199025 Chr.8: 11424320-11424320 on Build GRCh38
rs753301707 Chr.8: 11424403-11424403 on Build GRCh38
rs761572595 Chr.8: 11424394-11424394 on Build GRCh38
rs199968652 Chr.8: 11424392-11424392 on Build GRCh38
rs547076790 Chr.8: 11424336-11424336 on Build GRCh38
rs368459570 Chr.8: 11424417-11424417 on Build GRCh38
rs984126198 Chr.8: 11424448-11424448 on Build GRCh38
rs370416023 Chr.8: 11424354-11424354 on Build GRCh38
rs764004616 Chr.8: 11424337-11424337 on Build GRCh38
rs202235860 Chr.8: 11424340-11424340 on Build GRCh38
rs777181659 Chr.8: 11424423-11424424 on Build GRCh38
rs775453862 Chr.8: 11424398-11424398 on Build GRCh38
rs759963618 Chr.8: 11424450-11424452 on Build GRCh38
rs368537961 Chr.8: 11424342-11424342 on Build GRCh38
rs572193573 Chr.8: 11424435-11424435 on Build GRCh38
rs764168254 Chr.8: 11424393-11424393 on Build GRCh38
rs770101595 Chr.8: 11424431-11424431 on Build GRCh38
rs376807405 Chr.8: 11424325-11424325 on Build GRCh38
rs768845937 Chr.8: 11424444-11424444 on Build GRCh38
rs373564873 Chr.8: 11424339-11424339 on Build GRCh38
rs927414203 Chr.8: 11424290-11424290 on Build GRCh38
rs555254008 Chr.8: 11424277-11424277 on Build GRCh38
rs996008684 Chr.8: 11424321-11424321 on Build GRCh38
rs761747530 Chr.8: 11424464-11424464 on Build GRCh38
rs771301144 Chr.8: 11424375-11424375 on Build GRCh38
rs143743025 Chr.8: 11424421-11424421 on Build GRCh38
rs1022321826 Chr.8: 11424351-11424351 on Build GRCh38
rs566681543 Chr.8: 11424395-11424395 on Build GRCh38
rs139424835 Chr.8: 11424455-11424455 on Build GRCh38
rs373612696 Chr.8: 11424341-11424341 on Build GRCh38
rs537742836 Chr.8: 11424330-11424330 on Build GRCh38
rs905824381 Chr.8: 11424253-11424253 on Build GRCh38
rs374639232 Chr.8: 11424409-11424409 on Build GRCh38
rs779476502 Chr.8: 11424350-11424350 on Build GRCh38
rs758850719 Chr.8: 11424355-11424355 on Build GRCh38
rs200511231 Chr.8: 11424328-11424328 on Build GRCh38
rs1044687619 Chr.8: 11424250-11424250 on Build GRCh38
rs754451449 Chr.8: 11424405-11424405 on Build GRCh38
rs765900981 Chr.8: 11424400-11424400 on Build GRCh38
rs769766563 Chr.8: 11424388-11424388 on Build GRCh38
rs1016067086 Chr.8: 11424318-11424318 on Build GRCh38
rs141292162 Chr.8: 11424418-11424418 on Build GRCh38
rs771780754 Chr.8: 11424465-11424465 on Build GRCh38
rs746097006 Chr.8: 11424387-11424387 on Build GRCh38
rs1057461198 Chr.8: 11424276-11424276 on Build GRCh38
rs763754784 Chr.8: 11424343-11424344 on Build GRCh38
rs1038192816 Chr.8: 11424440-11424440 on Build GRCh38
rs938768205 Chr.8: 11424301-11424301 on Build GRCh38
rs755649418 Chr.8: 11424347-11424347 on Build GRCh38
rs940760420 Chr.8: 11424434-11424434 on Build GRCh38
rs778299082 Chr.8: 11424408-11424408 on Build GRCh38
rs771433749 Chr.8: 11424378-11424381 on Build GRCh38
rs778137618 Chr.8: 11424367-11424367 on Build GRCh38
rs1002884082 Chr.8: 11424275-11424275 on Build GRCh38
rs746092041 Chr.8: 11424429-11424429 on Build GRCh38
rs370672901 Chr.8: 11424314-11424314 on Build GRCh38
rs761098784 Chr.8: 11424378-11424378 on Build GRCh38
rs1005107747 Chr.8: 11424284-11424284 on Build GRCh38

View Full Product Details