Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120626148-120626417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1GALT1C1
Entrez Gene ID
29071
Gene Name
C1GALT1 specific chaperone 1
Gene Aliases
C1GALT2, C38H2-L1, COSMC, HSPC067, MST143, TNPS
UniGene
Hs.643920
Gene Chromosome Location
Chr.X: 120625674-120630150 on Build GRCh38
Transcripts
NM_001011551.2 NM_152692.4

SNP Detail

SNP ID Chromosome Location
rs774151751 Chr.X: 120626284-120626284 on Build GRCh38
rs538301204 Chr.X: 120626391-120626391 on Build GRCh38
rs16995907 Chr.X: 120626176-120626176 on Build GRCh38
rs751436721 Chr.X: 120626193-120626193 on Build GRCh38
rs758689561 Chr.X: 120626379-120626379 on Build GRCh38
rs754099112 Chr.X: 120626338-120626338 on Build GRCh38
rs142004998 Chr.X: 120626187-120626187 on Build GRCh38
rs749131756 Chr.X: 120626227-120626227 on Build GRCh38
rs766957795 Chr.X: 120626332-120626332 on Build GRCh38
rs11556782 Chr.X: 120626222-120626222 on Build GRCh38
rs371736291 Chr.X: 120626340-120626340 on Build GRCh38
rs936090662 Chr.X: 120626320-120626320 on Build GRCh38
rs769812960 Chr.X: 120626204-120626204 on Build GRCh38
rs765074731 Chr.X: 120626376-120626376 on Build GRCh38
rs367863648 Chr.X: 120626315-120626315 on Build GRCh38
rs772845587 Chr.X: 120626330-120626330 on Build GRCh38
rs868477895 Chr.X: 120626194-120626194 on Build GRCh38
rs758473662 Chr.X: 120626183-120626183 on Build GRCh38
rs768384820 Chr.X: 120626283-120626283 on Build GRCh38
rs761350200 Chr.X: 120626301-120626301 on Build GRCh38
rs775128621 Chr.X: 120626217-120626217 on Build GRCh38
rs754085776 Chr.X: 120626174-120626174 on Build GRCh38
rs752925954 Chr.X: 120626358-120626358 on Build GRCh38
rs748182662 Chr.X: 120626177-120626177 on Build GRCh38
rs777803355 Chr.X: 120626384-120626384 on Build GRCh38
rs371041040 Chr.X: 120626246-120626246 on Build GRCh38

View Full Product Details