Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47577037-47577308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSPG5
Entrez Gene ID
10675
Gene Name
chondroitin sulfate proteoglycan 5
Gene Aliases
NGC
UniGene
Hs.45127
Gene Chromosome Location
Chr.3: 47562238-47580240 on Build GRCh38
Transcripts
NM_001206942.1 NM_001206943.1 NM_001206944.1 NM_001206945.1 NM_006574.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4149940 c.790G>C p.D264H Chr.3: 47577236-47577236 on Build GRCh38
5765210 c.732G>A p.E244E Chr.3: 47577294-47577294 on Build GRCh38
6364474 c.772A>G p.T258A Chr.3: 47577254-47577254 on Build GRCh38
6883039 c.989G>A p.G330E Chr.3: 47577037-47577037 on Build GRCh38
3594790 c.822C>T p.D274D Chr.3: 47577204-47577204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941296028 Chr.3: 47577185-47577185 on Build GRCh38
rs748645809 Chr.3: 47577063-47577063 on Build GRCh38
rs778125605 Chr.3: 47577199-47577199 on Build GRCh38
rs774101025 Chr.3: 47577263-47577263 on Build GRCh38
rs1037972886 Chr.3: 47577216-47577216 on Build GRCh38
rs753681401 Chr.3: 47577136-47577136 on Build GRCh38
rs143267577 Chr.3: 47577182-47577182 on Build GRCh38
rs771673986 Chr.3: 47577267-47577267 on Build GRCh38
rs781450382 Chr.3: 47577165-47577165 on Build GRCh38
rs770381855 Chr.3: 47577082-47577082 on Build GRCh38
rs983265672 Chr.3: 47577088-47577088 on Build GRCh38
rs755159658 Chr.3: 47577164-47577164 on Build GRCh38
rs917340110 Chr.3: 47577155-47577155 on Build GRCh38
rs773713041 Chr.3: 47577089-47577089 on Build GRCh38
rs759131931 Chr.3: 47577266-47577266 on Build GRCh38
rs201571337 Chr.3: 47577103-47577103 on Build GRCh38
rs752897563 Chr.3: 47577258-47577258 on Build GRCh38
rs557218610 Chr.3: 47577149-47577149 on Build GRCh38
rs773870725 Chr.3: 47577100-47577100 on Build GRCh38
rs1046971972 Chr.3: 47577134-47577134 on Build GRCh38
rs1044409047 Chr.3: 47577213-47577213 on Build GRCh38
rs755004660 Chr.3: 47577057-47577057 on Build GRCh38
rs751703859 Chr.3: 47577154-47577154 on Build GRCh38
rs763952914 Chr.3: 47577128-47577128 on Build GRCh38
rs936280211 Chr.3: 47577253-47577253 on Build GRCh38
rs749728042 Chr.3: 47577202-47577202 on Build GRCh38
rs771167725 Chr.3: 47577210-47577210 on Build GRCh38
rs926194510 Chr.3: 47577242-47577242 on Build GRCh38
rs147289456 Chr.3: 47577111-47577111 on Build GRCh38
rs775350599 Chr.3: 47577180-47577180 on Build GRCh38
rs774986935 Chr.3: 47577279-47577279 on Build GRCh38
rs748169326 Chr.3: 47577173-47577173 on Build GRCh38
rs922413792 Chr.3: 47577101-47577101 on Build GRCh38
rs760639148 Chr.3: 47577117-47577117 on Build GRCh38

View Full Product Details