Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 224380011-224380242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM124B
Entrez Gene ID
79843
Gene Name
family with sequence similarity 124 member B
Gene Aliases
-
UniGene
Hs.147585
Gene Chromosome Location
Chr.2: 224378698-224401994 on Build GRCh38
Transcripts
NM_001122779.1 NM_024785.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3046439 c.859G>A p.G287S Chr.2: 224380082-224380082 on Build GRCh38
6883223 c.869C>T p.S290F Chr.2: 224380072-224380072 on Build GRCh38
6500339 c.894T>G p.P298P Chr.2: 224380047-224380047 on Build GRCh38
5606532 c.856C>T p.Q286* Chr.2: 224380085-224380085 on Build GRCh38
4851581 c.886G>C p.E296Q Chr.2: 224380055-224380055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972144765 Chr.2: 224380049-224380049 on Build GRCh38
rs3738953 Chr.2: 224380206-224380206 on Build GRCh38
rs771925196 Chr.2: 224380119-224380119 on Build GRCh38
rs546571913 Chr.2: 224380043-224380043 on Build GRCh38
rs939568122 Chr.2: 224380177-224380177 on Build GRCh38
rs745313047 Chr.2: 224380080-224380080 on Build GRCh38
rs867913272 Chr.2: 224380159-224380159 on Build GRCh38
rs941727283 Chr.2: 224380146-224380146 on Build GRCh38
rs887415107 Chr.2: 224380117-224380117 on Build GRCh38
rs138115641 Chr.2: 224380039-224380039 on Build GRCh38
rs769418312 Chr.2: 224380097-224380097 on Build GRCh38
rs773964645 Chr.2: 224380127-224380127 on Build GRCh38
rs1039012406 Chr.2: 224380165-224380165 on Build GRCh38
rs994336856 Chr.2: 224380176-224380176 on Build GRCh38
rs3738954 Chr.2: 224380171-224380171 on Build GRCh38
rs920787867 Chr.2: 224380058-224380058 on Build GRCh38
rs74519948 Chr.2: 224380185-224380185 on Build GRCh38
rs753965262 Chr.2: 224380175-224380175 on Build GRCh38
rs541647437 Chr.2: 224380031-224380031 on Build GRCh38
rs752966609 Chr.2: 224380209-224380209 on Build GRCh38
rs761839956 Chr.2: 224380167-224380167 on Build GRCh38
rs775352641 Chr.2: 224380103-224380103 on Build GRCh38
rs764025283 Chr.2: 224380122-224380122 on Build GRCh38
rs143597754 Chr.2: 224380111-224380111 on Build GRCh38
rs1035149311 Chr.2: 224380182-224380182 on Build GRCh38
rs200510905 Chr.2: 224380155-224380155 on Build GRCh38
rs952194895 Chr.2: 224380098-224380098 on Build GRCh38

View Full Product Details