Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60868479-60868752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
UniGene
Hs.655028
Gene Chromosome Location
Chr.17: 60677811-61392838 on Build GRCh38
Transcripts
NM_001099432.1 NM_001320470.1 NM_017679.3 XM_005257472.1 XM_011524939.1 XM_011524940.1 XM_011524941.1 XM_011524942.2 XM_011524943.2 XM_011524944.1 XM_011524945.2 XM_011524946.2 XM_011524947.1 XM_011524948.2 XM_011524949.2 XM_011524950.1 XM_011524951.2 XM_017024783.1 XM_017024784.1 XM_017024785.1 XM_017024786.1 XM_017024787.1 XM_017024788.1 XM_017024789.1 XM_017024790.1 XM_017024791.1 XM_017024792.1 XM_017024793.1 XM_017024794.1 XM_017024795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5005718 c.559A>G p.I187V Chr.17: 60868658-60868658 on Build GRCh38
1384964 c.583C>T p.R195W Chr.17: 60868682-60868682 on Build GRCh38
4151695 c.485C>A p.P162Q Chr.17: 60868584-60868584 on Build GRCh38
4636345 c.488C>T p.P163L Chr.17: 60868587-60868587 on Build GRCh38
4636346 c.488C>T p.P163L Chr.17: 60868587-60868587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185132789 Chr.17: 60868689-60868689 on Build GRCh38
rs769519972 Chr.17: 60868603-60868603 on Build GRCh38
rs771698366 Chr.17: 60868643-60868643 on Build GRCh38
rs773855608 Chr.17: 60868633-60868633 on Build GRCh38
rs762437454 Chr.17: 60868683-60868683 on Build GRCh38
rs377752373 Chr.17: 60868682-60868682 on Build GRCh38
rs746042915 Chr.17: 60868576-60868576 on Build GRCh38
rs949430011 Chr.17: 60868620-60868620 on Build GRCh38
rs764655566 Chr.17: 60868545-60868545 on Build GRCh38
rs936622349 Chr.17: 60868569-60868569 on Build GRCh38
rs1029617080 Chr.17: 60868666-60868666 on Build GRCh38
rs542393076 Chr.17: 60868724-60868724 on Build GRCh38
rs534209124 Chr.17: 60868539-60868539 on Build GRCh38
rs745684704 Chr.17: 60868561-60868561 on Build GRCh38
rs765925789 Chr.17: 60868527-60868527 on Build GRCh38
rs751781813 Chr.17: 60868725-60868725 on Build GRCh38
rs73320618 Chr.17: 60868588-60868588 on Build GRCh38
rs767677837 Chr.17: 60868513-60868513 on Build GRCh38
rs1037897093 Chr.17: 60868698-60868698 on Build GRCh38
rs757389896 Chr.17: 60868567-60868567 on Build GRCh38
rs574345303 Chr.17: 60868538-60868538 on Build GRCh38
rs761069193 Chr.17: 60868675-60868675 on Build GRCh38
rs777460667 Chr.17: 60868727-60868727 on Build GRCh38
rs770388498 Chr.17: 60868630-60868630 on Build GRCh38
rs759133042 Chr.17: 60868640-60868640 on Build GRCh38
rs773183785 Chr.17: 60868616-60868616 on Build GRCh38
rs925186081 Chr.17: 60868562-60868562 on Build GRCh38
rs867427251 Chr.17: 60868729-60868729 on Build GRCh38
rs759218597 Chr.17: 60868537-60868537 on Build GRCh38
rs756492551 Chr.17: 60868544-60868544 on Build GRCh38
rs142836169 Chr.17: 60868560-60868560 on Build GRCh38
rs151035589 Chr.17: 60868730-60868730 on Build GRCh38
rs775069588 Chr.17: 60868645-60868645 on Build GRCh38
rs758696075 Chr.17: 60868587-60868587 on Build GRCh38
rs758620599 Chr.17: 60868716-60868716 on Build GRCh38
rs188031327 Chr.17: 60868690-60868690 on Build GRCh38
rs764414602 Chr.17: 60868679-60868679 on Build GRCh38
rs773945154 Chr.17: 60868536-60868536 on Build GRCh38
rs749209369 Chr.17: 60868617-60868617 on Build GRCh38
rs747973805 Chr.17: 60868593-60868593 on Build GRCh38
rs754342410 Chr.17: 60868549-60868549 on Build GRCh38
rs779152948 Chr.17: 60868574-60868574 on Build GRCh38

View Full Product Details