Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67236205-67236464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAGAB
Entrez Gene ID
79719
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
KPPP1, PPKP1, PPKP1A, p34
UniGene
Hs.254642
Gene Chromosome Location
Chr.15: 67200675-67255198 on Build GRCh38
Transcripts
NM_001271885.1 NM_001271886.1 NM_024666.4 XM_011522020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
401570 c.355G>C p.E119Q Chr.15: 67236414-67236414 on Build GRCh38
4056429 c.338T>C p.V113A Chr.15: 67236431-67236431 on Build GRCh38
964272 c.342C>T p.C114C Chr.15: 67236427-67236427 on Build GRCh38
6464039 c.329T>A p.M110K Chr.15: 67236440-67236440 on Build GRCh38
6349259 c.320C>A p.P107H Chr.15: 67236449-67236449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983590733 Chr.15: 67236246-67236246 on Build GRCh38
rs758889595 Chr.15: 67236402-67236402 on Build GRCh38
rs35008448 Chr.15: 67236239-67236239 on Build GRCh38
rs767024623 Chr.15: 67236410-67236410 on Build GRCh38
rs772914150 Chr.15: 67236350-67236350 on Build GRCh38
rs186662479 Chr.15: 67236440-67236440 on Build GRCh38
rs16950788 Chr.15: 67236314-67236314 on Build GRCh38
rs895673292 Chr.15: 67236391-67236391 on Build GRCh38
rs912082600 Chr.15: 67236302-67236302 on Build GRCh38
rs775172947 Chr.15: 67236379-67236379 on Build GRCh38
rs375202846 Chr.15: 67236247-67236248 on Build GRCh38
rs776580912 Chr.15: 67236267-67236267 on Build GRCh38
rs182392210 Chr.15: 67236384-67236384 on Build GRCh38
rs763590567 Chr.15: 67236394-67236394 on Build GRCh38
rs760548483 Chr.15: 67236432-67236432 on Build GRCh38
rs749184555 Chr.15: 67236357-67236357 on Build GRCh38
rs371926366 Chr.15: 67236426-67236426 on Build GRCh38
rs893854724 Chr.15: 67236411-67236411 on Build GRCh38
rs561525955 Chr.15: 67236246-67236249 on Build GRCh38
rs768513159 Chr.15: 67236258-67236258 on Build GRCh38
rs964348470 Chr.15: 67236234-67236234 on Build GRCh38
rs759361906 Chr.15: 67236375-67236375 on Build GRCh38
rs974416499 Chr.15: 67236239-67236239 on Build GRCh38
rs755439411 Chr.15: 67236416-67236416 on Build GRCh38
rs759156533 Chr.15: 67236377-67236378 on Build GRCh38
rs865986004 Chr.15: 67236414-67236414 on Build GRCh38
rs946756886 Chr.15: 67236327-67236327 on Build GRCh38
rs930391607 Chr.15: 67236277-67236277 on Build GRCh38
rs908694678 Chr.15: 67236306-67236306 on Build GRCh38
rs565607839 Chr.15: 67236279-67236279 on Build GRCh38
rs767126385 Chr.15: 67236377-67236377 on Build GRCh38
rs770872438 Chr.15: 67236360-67236360 on Build GRCh38
rs200664620 Chr.15: 67236373-67236373 on Build GRCh38
rs545605172 Chr.15: 67236235-67236235 on Build GRCh38
rs756296586 Chr.15: 67236433-67236433 on Build GRCh38
rs769442825 Chr.15: 67236295-67236295 on Build GRCh38
rs1055058820 Chr.15: 67236405-67236405 on Build GRCh38
rs1044353187 Chr.15: 67236427-67236427 on Build GRCh38
rs191746737 Chr.15: 67236335-67236335 on Build GRCh38
rs748466481 Chr.15: 67236430-67236430 on Build GRCh38
rs774087773 Chr.15: 67236363-67236363 on Build GRCh38
rs920308033 Chr.15: 67236247-67236247 on Build GRCh38
rs771975734 Chr.15: 67236422-67236423 on Build GRCh38
rs751937153 Chr.15: 67236412-67236412 on Build GRCh38
rs975053797 Chr.15: 67236312-67236312 on Build GRCh38
rs775155034 Chr.15: 67236332-67236332 on Build GRCh38
rs753300689 Chr.15: 67236400-67236400 on Build GRCh38
rs760092902 Chr.15: 67236390-67236390 on Build GRCh38
rs775489702 Chr.15: 67236425-67236425 on Build GRCh38

View Full Product Details