Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113428959-113429192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADPRHL1
Entrez Gene ID
113622
Gene Name
ADP-ribosylhydrolase like 1
Gene Aliases
ARH2
UniGene
Hs.98669
Gene Chromosome Location
Chr.13: 113421940-113453524 on Build GRCh38
Transcripts
NM_138430.4 NM_199162.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4166698 c.634C>T p.R212W Chr.13: 113428964-113428964 on Build GRCh38
5550883 c.620G>C p.C207S Chr.13: 113428978-113428978 on Build GRCh38
3467412 c.514G>A p.G172S Chr.13: 113429084-113429084 on Build GRCh38
4503046 c.629C>T p.T210I Chr.13: 113428969-113428969 on Build GRCh38
3711006 c.506-1G>C p.? Chr.13: 113429093-113429093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148154052 Chr.13: 113429106-113429107 on Build GRCh38
rs181386121 Chr.13: 113429002-113429002 on Build GRCh38
rs1050676645 Chr.13: 113429017-113429017 on Build GRCh38
rs746049429 Chr.13: 113429141-113429141 on Build GRCh38
rs767237977 Chr.13: 113429128-113429174 on Build GRCh38
rs758943911 Chr.13: 113429127-113429145 on Build GRCh38
rs1022771361 Chr.13: 113429082-113429082 on Build GRCh38
rs537663983 Chr.13: 113429154-113429154 on Build GRCh38
rs139697294 Chr.13: 113429015-113429015 on Build GRCh38
rs142661838 Chr.13: 113428978-113428978 on Build GRCh38
rs762877803 Chr.13: 113429108-113429109 on Build GRCh38
rs199692255 Chr.13: 113429005-113429005 on Build GRCh38
rs756057986 Chr.13: 113429119-113429119 on Build GRCh38
rs956081591 Chr.13: 113429135-113429135 on Build GRCh38
rs932206858 Chr.13: 113429016-113429016 on Build GRCh38
rs752686820 Chr.13: 113429117-113429117 on Build GRCh38
rs758114074 Chr.13: 113429022-113429022 on Build GRCh38
rs773462863 Chr.13: 113428994-113428994 on Build GRCh38
rs970126707 Chr.13: 113429092-113429092 on Build GRCh38
rs756251846 Chr.13: 113429035-113429035 on Build GRCh38
rs764812133 Chr.13: 113429018-113429018 on Build GRCh38
rs771184612 Chr.13: 113429052-113429052 on Build GRCh38
rs567783735 Chr.13: 113429096-113429096 on Build GRCh38
rs748045770 Chr.13: 113429029-113429029 on Build GRCh38
rs551989043 Chr.13: 113429115-113429115 on Build GRCh38
rs772299702 Chr.13: 113429057-113429057 on Build GRCh38
rs746264712 Chr.13: 113429056-113429056 on Build GRCh38
rs753094234 Chr.13: 113429011-113429011 on Build GRCh38
rs559793322 Chr.13: 113429070-113429070 on Build GRCh38
rs749397359 Chr.13: 113429129-113429129 on Build GRCh38
rs907962027 Chr.13: 113428989-113428989 on Build GRCh38
rs10700715 Chr.13: 113429114-113429115 on Build GRCh38
rs769603778 Chr.13: 113429037-113429041 on Build GRCh38
rs920619446 Chr.13: 113429013-113429013 on Build GRCh38
rs763157942 Chr.13: 113429000-113429000 on Build GRCh38
rs371620550 Chr.13: 113429155-113429155 on Build GRCh38
rs386380796 Chr.13: 113429113-113429114 on Build GRCh38
rs750172361 Chr.13: 113429020-113429020 on Build GRCh38
rs377658823 Chr.13: 113429048-113429048 on Build GRCh38
rs368103191 Chr.13: 113429138-113429138 on Build GRCh38
rs190623578 Chr.13: 113428976-113428976 on Build GRCh38
rs757232254 Chr.13: 113429134-113429134 on Build GRCh38
rs748290770 Chr.13: 113428979-113428979 on Build GRCh38
rs74520894 Chr.13: 113429158-113429158 on Build GRCh38
rs769817799 Chr.13: 113428983-113428983 on Build GRCh38
rs751125228 Chr.13: 113429099-113429099 on Build GRCh38
rs754691717 Chr.13: 113429023-113429023 on Build GRCh38
rs981519335 Chr.13: 113429100-113429100 on Build GRCh38
rs547476358 Chr.13: 113429066-113429066 on Build GRCh38
rs370545967 Chr.13: 113429055-113429055 on Build GRCh38
rs9635114 Chr.13: 113429001-113429001 on Build GRCh38
rs375108772 Chr.13: 113429006-113429006 on Build GRCh38
rs533507871 Chr.13: 113429071-113429071 on Build GRCh38
rs1009069798 Chr.13: 113429044-113429044 on Build GRCh38
rs766086834 Chr.13: 113429098-113429098 on Build GRCh38
rs902922489 Chr.13: 113429051-113429051 on Build GRCh38
rs772865696 Chr.13: 113429090-113429090 on Build GRCh38
rs777619526 Chr.13: 113429125-113429125 on Build GRCh38
rs767210032 Chr.13: 113429116-113429116 on Build GRCh38
rs749307877 Chr.13: 113429049-113429049 on Build GRCh38
rs760849716 Chr.13: 113428992-113428993 on Build GRCh38

View Full Product Details