Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 13906273-13906520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETV1
Entrez Gene ID
2115
Gene Name
ETS variant 1
Gene Aliases
ER81
UniGene
Hs.22634
Gene Chromosome Location
Chr.7: 13891229-13991425 on Build GRCh38
Transcripts
NM_001163147.1 NM_001163148.1 NM_001163149.1 NM_001163150.1 NM_001163151.1 NM_001163152.1 NM_004956.4 NR_120445.1 XM_005249635.4 XM_005249636.4 XM_011515167.2 XM_011515168.2 XM_011515170.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
452448 c.1105G>C p.E369Q Chr.7: 13906435-13906435 on Build GRCh38
3634134 c.1073G>A p.G358D Chr.7: 13906467-13906467 on Build GRCh38
1086160 c.1075C>T p.R359* Chr.7: 13906465-13906465 on Build GRCh38
5935267 c.1036G>A p.D346N Chr.7: 13906504-13906504 on Build GRCh38
6930707 c.1079G>A p.G360D Chr.7: 13906461-13906461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767110553 Chr.7: 13906468-13906468 on Build GRCh38
rs551638033 Chr.7: 13906364-13906364 on Build GRCh38
rs763791524 Chr.7: 13906421-13906421 on Build GRCh38
rs184679518 Chr.7: 13906402-13906402 on Build GRCh38
rs6942760 Chr.7: 13906338-13906338 on Build GRCh38
rs752385952 Chr.7: 13906381-13906385 on Build GRCh38
rs371617913 Chr.7: 13906398-13906398 on Build GRCh38
rs758040765 Chr.7: 13906382-13906383 on Build GRCh38
rs757006753 Chr.7: 13906457-13906457 on Build GRCh38
rs372949158 Chr.7: 13906439-13906439 on Build GRCh38
rs113241897 Chr.7: 13906306-13906306 on Build GRCh38
rs937462195 Chr.7: 13906372-13906372 on Build GRCh38
rs950088033 Chr.7: 13906327-13906327 on Build GRCh38
rs891005056 Chr.7: 13906325-13906325 on Build GRCh38
rs567326719 Chr.7: 13906477-13906477 on Build GRCh38
rs1027136314 Chr.7: 13906475-13906475 on Build GRCh38
rs775856362 Chr.7: 13906405-13906405 on Build GRCh38
rs749957333 Chr.7: 13906471-13906471 on Build GRCh38
rs763238437 Chr.7: 13906420-13906420 on Build GRCh38
rs149316422 Chr.7: 13906354-13906354 on Build GRCh38
rs374550516 Chr.7: 13906401-13906401 on Build GRCh38
rs753613347 Chr.7: 13906498-13906498 on Build GRCh38

View Full Product Details