Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27138017-27138264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR30
Entrez Gene ID
339779
Gene Name
proline rich 30
Gene Aliases
C2orf53
UniGene
Hs.136555
Gene Chromosome Location
Chr.2: 27136847-27139464 on Build GRCh38
Transcripts
NM_178553.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5862316 c.126G>T p.Q42H Chr.2: 27138204-27138204 on Build GRCh38
347240 c.134C>A p.P45H Chr.2: 27138196-27138196 on Build GRCh38
7318946 c.232T>A p.S78T Chr.2: 27138098-27138098 on Build GRCh38
5695122 c.94C>T p.P32S Chr.2: 27138236-27138236 on Build GRCh38
3580523 c.177C>T p.P59P Chr.2: 27138153-27138153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141290844 Chr.2: 27138180-27138180 on Build GRCh38
rs766275352 Chr.2: 27138117-27138117 on Build GRCh38
rs201906024 Chr.2: 27138098-27138098 on Build GRCh38
rs753946403 Chr.2: 27138043-27138043 on Build GRCh38
rs113380452 Chr.2: 27138111-27138111 on Build GRCh38
rs79201461 Chr.2: 27138059-27138059 on Build GRCh38
rs556394185 Chr.2: 27138127-27138127 on Build GRCh38
rs550077691 Chr.2: 27138134-27138134 on Build GRCh38
rs780283996 Chr.2: 27138091-27138091 on Build GRCh38
rs570671962 Chr.2: 27138161-27138161 on Build GRCh38
rs113634430 Chr.2: 27138154-27138154 on Build GRCh38
rs747409237 Chr.2: 27138172-27138172 on Build GRCh38
rs754108199 Chr.2: 27138148-27138148 on Build GRCh38
rs144004044 Chr.2: 27138097-27138097 on Build GRCh38
rs144158821 Chr.2: 27138201-27138201 on Build GRCh38
rs149337790 Chr.2: 27138160-27138160 on Build GRCh38
rs1056922750 Chr.2: 27138177-27138177 on Build GRCh38
rs759443606 Chr.2: 27138206-27138206 on Build GRCh38
rs553610498 Chr.2: 27138158-27138158 on Build GRCh38
rs140556985 Chr.2: 27138110-27138110 on Build GRCh38
rs764452109 Chr.2: 27138184-27138184 on Build GRCh38
rs779027857 Chr.2: 27138072-27138072 on Build GRCh38
rs370650817 Chr.2: 27138082-27138082 on Build GRCh38
rs371357819 Chr.2: 27138188-27138188 on Build GRCh38
rs776384065 Chr.2: 27138100-27138100 on Build GRCh38
rs767357441 Chr.2: 27138211-27138211 on Build GRCh38
rs762025138 Chr.2: 27138221-27138221 on Build GRCh38
rs538214448 Chr.2: 27138157-27138157 on Build GRCh38
rs754132727 Chr.2: 27138197-27138198 on Build GRCh38
rs201152841 Chr.2: 27138193-27138193 on Build GRCh38
rs149656537 Chr.2: 27138212-27138212 on Build GRCh38
rs140765935 Chr.2: 27138105-27138105 on Build GRCh38
rs75770831 Chr.2: 27138130-27138130 on Build GRCh38
rs755435255 Chr.2: 27138178-27138178 on Build GRCh38
rs367785900 Chr.2: 27138076-27138076 on Build GRCh38
rs61739485 Chr.2: 27138181-27138181 on Build GRCh38
rs757790492 Chr.2: 27138208-27138209 on Build GRCh38
rs774382660 Chr.2: 27138202-27138202 on Build GRCh38
rs566922623 Chr.2: 27138074-27138074 on Build GRCh38
rs538148113 Chr.2: 27138115-27138115 on Build GRCh38
rs765440658 Chr.2: 27138224-27138224 on Build GRCh38

View Full Product Details