Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 6851525-6851750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMPK2
Entrez Gene ID
129607
Gene Name
cytidine/uridine monophosphate kinase 2
Gene Aliases
NDK, TMPK2, TYKi, UMP-CMPK2
UniGene
Hs.7155
Gene Chromosome Location
Chr.2: 6840417-6866635 on Build GRCh38
Transcripts
NM_001256477.1 NM_001256478.1 NM_207315.3 NR_046236.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3056175 c.1072C>T p.P358S Chr.2: 6851604-6851604 on Build GRCh38
3910519 c.1140G>A p.E380E Chr.2: 6851536-6851536 on Build GRCh38
3056176 c.1014C>A p.T338T Chr.2: 6851662-6851662 on Build GRCh38
3963006 c.1008G>T p.T336T Chr.2: 6851668-6851668 on Build GRCh38
1022445 c.1111C>A p.L371I Chr.2: 6851565-6851565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780750427 Chr.2: 6851722-6851722 on Build GRCh38
rs756085131 Chr.2: 6851547-6851547 on Build GRCh38
rs773552908 Chr.2: 6851638-6851638 on Build GRCh38
rs763211338 Chr.2: 6851651-6851651 on Build GRCh38
rs1054542683 Chr.2: 6851715-6851715 on Build GRCh38
rs200083396 Chr.2: 6851628-6851628 on Build GRCh38
rs770125101 Chr.2: 6851636-6851636 on Build GRCh38
rs778629159 Chr.2: 6851581-6851581 on Build GRCh38
rs1032083814 Chr.2: 6851660-6851660 on Build GRCh38
rs746481321 Chr.2: 6851604-6851604 on Build GRCh38
rs748552102 Chr.2: 6851635-6851635 on Build GRCh38
rs764995155 Chr.2: 6851686-6851686 on Build GRCh38
rs201507613 Chr.2: 6851605-6851605 on Build GRCh38
rs753687740 Chr.2: 6851566-6851566 on Build GRCh38
rs774445534 Chr.2: 6851658-6851658 on Build GRCh38
rs1049393398 Chr.2: 6851676-6851676 on Build GRCh38
rs374669579 Chr.2: 6851623-6851623 on Build GRCh38
rs927335351 Chr.2: 6851549-6851549 on Build GRCh38
rs889492848 Chr.2: 6851681-6851681 on Build GRCh38
rs983918116 Chr.2: 6851577-6851577 on Build GRCh38
rs759561949 Chr.2: 6851663-6851663 on Build GRCh38
rs753647983 Chr.2: 6851669-6851669 on Build GRCh38
rs760812828 Chr.2: 6851695-6851695 on Build GRCh38
rs757211308 Chr.2: 6851680-6851680 on Build GRCh38
rs536045288 Chr.2: 6851700-6851700 on Build GRCh38
rs756943671 Chr.2: 6851570-6851570 on Build GRCh38
rs750217559 Chr.2: 6851690-6851690 on Build GRCh38
rs754383037 Chr.2: 6851709-6851709 on Build GRCh38
rs758107640 Chr.2: 6851583-6851583 on Build GRCh38
rs372012904 Chr.2: 6851668-6851668 on Build GRCh38
rs745580649 Chr.2: 6851582-6851582 on Build GRCh38
rs763898811 Chr.2: 6851548-6851548 on Build GRCh38
rs768952552 Chr.2: 6851706-6851706 on Build GRCh38
rs965082360 Chr.2: 6851716-6851716 on Build GRCh38
rs376917956 Chr.2: 6851704-6851704 on Build GRCh38
rs909756124 Chr.2: 6851616-6851616 on Build GRCh38
rs367752153 Chr.2: 6851655-6851655 on Build GRCh38
rs372816766 Chr.2: 6851589-6851589 on Build GRCh38

View Full Product Details