Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32215327-32215600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCDC2B
Entrez Gene ID
149069
Gene Name
doublecortin domain containing 2B
Gene Aliases
-
UniGene
Hs.424588
Gene Chromosome Location
Chr.1: 32208813-32216196 on Build GRCh38
Transcripts
NM_001099434.1 NR_133635.1 NR_133636.1 XM_011541795.2 XM_017001812.1 XM_017001813.1 XM_017001814.1 XM_017001815.1 XM_017001816.1 XM_017001817.1 XM_017001818.1 XM_017001819.1 XM_017001820.1 XM_017001821.1 XM_017001822.1 XM_017001823.1 XM_017001826.1 XM_017001827.1 XM_017001828.1 XM_017001829.1 XM_017001830.1

Gene Symbol
TMEM234
Entrez Gene ID
56063
Gene Name
transmembrane protein 234
Gene Aliases
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7
UniGene
Hs.272299
Gene Chromosome Location
Chr.1: 32204807-32222371 on Build GRCh38
Transcripts
NM_001099434.1 NR_133635.1 NR_133636.1 XM_011541795.2 XM_017001812.1 XM_017001813.1 XM_017001814.1 XM_017001815.1 XM_017001816.1 XM_017001817.1 XM_017001818.1 XM_017001819.1 XM_017001820.1 XM_017001821.1 XM_017001822.1 XM_017001823.1 XM_017001826.1 XM_017001827.1 XM_017001828.1 XM_017001829.1 XM_017001830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4850352 c.916G>C p.D306H Chr.1: 32215505-32215505 on Build GRCh38
5529592 c.922G>A p.E308K Chr.1: 32215511-32215511 on Build GRCh38
908270 c.942G>A p.E314E Chr.1: 32215531-32215531 on Build GRCh38
6905026 c.943C>A p.P315T Chr.1: 32215532-32215532 on Build GRCh38
5415030 c.879C>A p.H293Q Chr.1: 32215468-32215468 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1053279213 Chr.1: 32215381-32215381 on Build GRCh38
rs913360821 Chr.1: 32215445-32215445 on Build GRCh38
rs935103330 Chr.1: 32215379-32215379 on Build GRCh38
rs770809156 Chr.1: 32215361-32215361 on Build GRCh38
rs375062766 Chr.1: 32215470-32215470 on Build GRCh38
rs148605394 Chr.1: 32215408-32215408 on Build GRCh38
rs777619000 Chr.1: 32215501-32215501 on Build GRCh38
rs777457323 Chr.1: 32215579-32215579 on Build GRCh38
rs756519317 Chr.1: 32215482-32215483 on Build GRCh38
rs769557918 Chr.1: 32215498-32215498 on Build GRCh38
rs950081627 Chr.1: 32215571-32215571 on Build GRCh38
rs769582327 Chr.1: 32215449-32215449 on Build GRCh38
rs780508572 Chr.1: 32215495-32215495 on Build GRCh38
rs369087066 Chr.1: 32215474-32215474 on Build GRCh38
rs977012866 Chr.1: 32215429-32215429 on Build GRCh38
rs370546533 Chr.1: 32215575-32215575 on Build GRCh38
rs373546059 Chr.1: 32215490-32215490 on Build GRCh38
rs749060689 Chr.1: 32215508-32215508 on Build GRCh38
rs763264271 Chr.1: 32215555-32215555 on Build GRCh38
rs564374739 Chr.1: 32215491-32215491 on Build GRCh38
rs768087456 Chr.1: 32215410-32215410 on Build GRCh38
rs767355275 Chr.1: 32215548-32215551 on Build GRCh38
rs755371601 Chr.1: 32215560-32215560 on Build GRCh38
rs765507964 Chr.1: 32215484-32215484 on Build GRCh38
rs753969398 Chr.1: 32215477-32215477 on Build GRCh38
rs375054940 Chr.1: 32215463-32215463 on Build GRCh38
rs775191406 Chr.1: 32215485-32215485 on Build GRCh38
rs755727615 Chr.1: 32215576-32215576 on Build GRCh38
rs748901331 Chr.1: 32215580-32215580 on Build GRCh38
rs778367462 Chr.1: 32215484-32215489 on Build GRCh38
rs770593003 Chr.1: 32215516-32215516 on Build GRCh38
rs1013850068 Chr.1: 32215578-32215578 on Build GRCh38
rs761935416 Chr.1: 32215531-32215531 on Build GRCh38
rs35461680 Chr.1: 32215468-32215469 on Build GRCh38
rs144910030 Chr.1: 32215347-32215347 on Build GRCh38
rs11590012 Chr.1: 32215468-32215468 on Build GRCh38
rs546171672 Chr.1: 32215486-32215486 on Build GRCh38
rs776234848 Chr.1: 32215427-32215427 on Build GRCh38
rs747664545 Chr.1: 32215496-32215496 on Build GRCh38
rs771581836 Chr.1: 32215471-32215471 on Build GRCh38
rs750679988 Chr.1: 32215552-32215552 on Build GRCh38
rs755530985 Chr.1: 32215492-32215492 on Build GRCh38
rs766603599 Chr.1: 32215557-32215557 on Build GRCh38
rs765420044 Chr.1: 32215548-32215548 on Build GRCh38
rs944880344 Chr.1: 32215453-32215453 on Build GRCh38
rs774227457 Chr.1: 32215517-32215517 on Build GRCh38
rs925011817 Chr.1: 32215350-32215350 on Build GRCh38
rs752233679 Chr.1: 32215572-32215572 on Build GRCh38
rs759338691 Chr.1: 32215518-32215518 on Build GRCh38
rs142243330 Chr.1: 32215528-32215528 on Build GRCh38
rs979110287 Chr.1: 32215349-32215349 on Build GRCh38
rs761959806 Chr.1: 32215481-32215481 on Build GRCh38
rs747754033 Chr.1: 32215439-32215439 on Build GRCh38
rs753109418 Chr.1: 32215473-32215473 on Build GRCh38
rs573967968 Chr.1: 32215558-32215558 on Build GRCh38
rs781755862 Chr.1: 32215563-32215563 on Build GRCh38
rs913541321 Chr.1: 32215360-32215360 on Build GRCh38

View Full Product Details