Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129085370-129085635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOLPP1
Entrez Gene ID
57171
Gene Name
dolichyldiphosphatase 1
Gene Aliases
LSFR2
UniGene
Hs.21701
Gene Chromosome Location
Chr.9: 129081104-129090438 on Build GRCh38
Transcripts
NM_001135917.1 NM_020438.4 NR_072983.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3328196 c.412G>A p.V138M Chr.9: 129085567-129085567 on Build GRCh38
7365310 c.412G>C p.V138L Chr.9: 129085567-129085567 on Build GRCh38
6873691 c.448G>A p.V150I Chr.9: 129085603-129085603 on Build GRCh38
1105813 c.433G>A p.A145T Chr.9: 129085588-129085588 on Build GRCh38
6865814 c.429C>T p.L143L Chr.9: 129085584-129085584 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs971801290 Chr.9: 129085436-129085436 on Build GRCh38
rs139256216 Chr.9: 129085567-129085567 on Build GRCh38
rs1003297268 Chr.9: 129085421-129085421 on Build GRCh38
rs1031046635 Chr.9: 129085431-129085431 on Build GRCh38
rs761270655 Chr.9: 129085572-129085572 on Build GRCh38
rs148259476 Chr.9: 129085479-129085479 on Build GRCh38
rs868508095 Chr.9: 129085413-129085413 on Build GRCh38
rs1016056015 Chr.9: 129085410-129085410 on Build GRCh38
rs749464782 Chr.9: 129085542-129085542 on Build GRCh38
rs981125489 Chr.9: 129085484-129085484 on Build GRCh38
rs370550987 Chr.9: 129085588-129085588 on Build GRCh38
rs547931668 Chr.9: 129085458-129085458 on Build GRCh38
rs150999812 Chr.9: 129085541-129085541 on Build GRCh38
rs369531690 Chr.9: 129085513-129085513 on Build GRCh38
rs1028620750 Chr.9: 129085614-129085614 on Build GRCh38
rs372491970 Chr.9: 129085599-129085599 on Build GRCh38
rs758080322 Chr.9: 129085503-129085503 on Build GRCh38
rs1023950260 Chr.9: 129085391-129085391 on Build GRCh38
rs768955439 Chr.9: 129085555-129085555 on Build GRCh38
rs970601540 Chr.9: 129085404-129085404 on Build GRCh38
rs781239127 Chr.9: 129085533-129085533 on Build GRCh38
rs200420096 Chr.9: 129085491-129085491 on Build GRCh38
rs975707047 Chr.9: 129085502-129085502 on Build GRCh38
rs768563315 Chr.9: 129085467-129085467 on Build GRCh38
rs534146722 Chr.9: 129085402-129085402 on Build GRCh38
rs766794929 Chr.9: 129085587-129085587 on Build GRCh38
rs774514828 Chr.9: 129085477-129085477 on Build GRCh38
rs773927315 Chr.9: 129085568-129085568 on Build GRCh38
rs764979782 Chr.9: 129085499-129085499 on Build GRCh38
rs761184448 Chr.9: 129085494-129085494 on Build GRCh38
rs374582267 Chr.9: 129085514-129085514 on Build GRCh38
rs774263388 Chr.9: 129085566-129085566 on Build GRCh38
rs915232411 Chr.9: 129085481-129085481 on Build GRCh38
rs558448826 Chr.9: 129085605-129085605 on Build GRCh38
rs752333676 Chr.9: 129085501-129085501 on Build GRCh38
rs745648840 Chr.9: 129085536-129085536 on Build GRCh38
rs777096917 Chr.9: 129085505-129085505 on Build GRCh38

View Full Product Details