Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43236523-43236796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENOX1
Entrez Gene ID
55068
Gene Name
ecto-NOX disulfide-thiol exchanger 1
Gene Aliases
CNOX, PIG38, bA64J21.1, cCNOX
UniGene
Hs.128258
Gene Chromosome Location
Chr.13: 43213525-43786980 on Build GRCh38
Transcripts
NM_001127615.1 NM_001242863.1 NM_017993.3 XM_005266439.3 XM_011535126.2 XM_011535127.2 XM_011535128.2 XM_011535132.2 XM_017020637.1 XM_017020638.1 XM_017020639.1 XM_017020640.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1941226 c.1652G>A p.R551Q Chr.13: 43236698-43236698 on Build GRCh38
3813859 c.1650C>T p.D550D Chr.13: 43236700-43236700 on Build GRCh38
5124434 c.1632T>C p.V544V Chr.13: 43236718-43236718 on Build GRCh38
216303 c.1642A>C p.N548H Chr.13: 43236708-43236708 on Build GRCh38
432408 c.1612G>T p.E538* Chr.13: 43236738-43236738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780270531 Chr.13: 43236768-43236768 on Build GRCh38
rs781226809 Chr.13: 43236609-43236609 on Build GRCh38
rs756350881 Chr.13: 43236756-43236756 on Build GRCh38
rs752480816 Chr.13: 43236594-43236594 on Build GRCh38
rs748473129 Chr.13: 43236642-43236642 on Build GRCh38
rs756970272 Chr.13: 43236674-43236674 on Build GRCh38
rs751380722 Chr.13: 43236745-43236745 on Build GRCh38
rs774845638 Chr.13: 43236639-43236639 on Build GRCh38
rs1037608097 Chr.13: 43236742-43236742 on Build GRCh38
rs754961167 Chr.13: 43236592-43236592 on Build GRCh38
rs375265573 Chr.13: 43236633-43236633 on Build GRCh38
rs894141639 Chr.13: 43236645-43236645 on Build GRCh38
rs765196544 Chr.13: 43236717-43236717 on Build GRCh38
rs995895586 Chr.13: 43236687-43236687 on Build GRCh38
rs150329674 Chr.13: 43236703-43236703 on Build GRCh38
rs1043663396 Chr.13: 43236623-43236623 on Build GRCh38
rs560032590 Chr.13: 43236732-43236732 on Build GRCh38
rs772510240 Chr.13: 43236669-43236669 on Build GRCh38
rs529596547 Chr.13: 43236668-43236668 on Build GRCh38
rs371666980 Chr.13: 43236698-43236698 on Build GRCh38
rs563804415 Chr.13: 43236635-43236635 on Build GRCh38
rs946638719 Chr.13: 43236565-43236565 on Build GRCh38
rs369595948 Chr.13: 43236680-43236680 on Build GRCh38
rs745574731 Chr.13: 43236762-43236762 on Build GRCh38
rs371476221 Chr.13: 43236613-43236613 on Build GRCh38
rs752709853 Chr.13: 43236731-43236731 on Build GRCh38
rs140224530 Chr.13: 43236673-43236673 on Build GRCh38
rs780181898 Chr.13: 43236622-43236622 on Build GRCh38
rs546594153 Chr.13: 43236661-43236661 on Build GRCh38
rs367895284 Chr.13: 43236593-43236593 on Build GRCh38
rs758215229 Chr.13: 43236596-43236596 on Build GRCh38
rs763922807 Chr.13: 43236743-43236743 on Build GRCh38

View Full Product Details