Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219174707-219174932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPPD1
Entrez Gene ID
27013
Gene Name
cyclin Pas1/PHO80 domain containing 1
Gene Aliases
C2orf24, CGI-57
UniGene
Hs.4973
Gene Chromosome Location
Chr.2: 219171897-219178174 on Build GRCh38
Transcripts
NM_001321389.1 NM_001321390.1 NM_001321391.1 NM_015680.5 NR_135628.1 NR_135629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5572344 c.404A>G p.Y135C Chr.2: 219174884-219174884 on Build GRCh38
3798657 c.403T>C p.Y135H Chr.2: 219174885-219174885 on Build GRCh38
5058683 c.469C>T p.P157S Chr.2: 219174819-219174819 on Build GRCh38
3838640 c.436G>A p.D146N Chr.2: 219174852-219174852 on Build GRCh38
1405574 c.468G>A p.V156V Chr.2: 219174820-219174820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767158581 Chr.2: 219174810-219174810 on Build GRCh38
rs781375675 Chr.2: 219174840-219174840 on Build GRCh38
rs766202955 Chr.2: 219174868-219174868 on Build GRCh38
rs753919640 Chr.2: 219174853-219174855 on Build GRCh38
rs760364225 Chr.2: 219174817-219174817 on Build GRCh38
rs150288895 Chr.2: 219174853-219174853 on Build GRCh38
rs776355868 Chr.2: 219174874-219174874 on Build GRCh38
rs557930253 Chr.2: 219174831-219174831 on Build GRCh38
rs774148764 Chr.2: 219174804-219174804 on Build GRCh38
rs747373067 Chr.2: 219174837-219174837 on Build GRCh38
rs376110739 Chr.2: 219174884-219174884 on Build GRCh38
rs769797305 Chr.2: 219174849-219174849 on Build GRCh38
rs767400089 Chr.2: 219174811-219174812 on Build GRCh38
rs764645008 Chr.2: 219174889-219174889 on Build GRCh38
rs774356157 Chr.2: 219174850-219174850 on Build GRCh38
rs754531078 Chr.2: 219174824-219174824 on Build GRCh38
rs577791189 Chr.2: 219174832-219174832 on Build GRCh38
rs757527114 Chr.2: 219174839-219174839 on Build GRCh38
rs758625585 Chr.2: 219174739-219174739 on Build GRCh38
rs753281188 Chr.2: 219174883-219174883 on Build GRCh38
rs182502513 Chr.2: 219174863-219174863 on Build GRCh38
rs768634077 Chr.2: 219174793-219174793 on Build GRCh38
rs751634975 Chr.2: 219174823-219174823 on Build GRCh38
rs757371658 Chr.2: 219174861-219174863 on Build GRCh38
rs746011264 Chr.2: 219174846-219174846 on Build GRCh38
rs747153200 Chr.2: 219174743-219174743 on Build GRCh38
rs373415129 Chr.2: 219174880-219174880 on Build GRCh38
rs151314580 Chr.2: 219174784-219174784 on Build GRCh38
rs765780977 Chr.2: 219174830-219174830 on Build GRCh38
rs778071168 Chr.2: 219174741-219174741 on Build GRCh38
rs772938514 Chr.2: 219174811-219174811 on Build GRCh38
rs761670049 Chr.2: 219174806-219174806 on Build GRCh38
rs771930595 Chr.2: 219174854-219174854 on Build GRCh38
rs370404660 Chr.2: 219174760-219174760 on Build GRCh38
rs751982944 Chr.2: 219174894-219174894 on Build GRCh38
rs200097602 Chr.2: 219174865-219174865 on Build GRCh38
rs199735062 Chr.2: 219174822-219174822 on Build GRCh38
rs201411225 Chr.2: 219174836-219174836 on Build GRCh38
rs758767449 Chr.2: 219174876-219174876 on Build GRCh38
rs878915142 Chr.2: 219174900-219174900 on Build GRCh38
rs1044108580 Chr.2: 219174742-219174742 on Build GRCh38
rs745928930 Chr.2: 219174785-219174785 on Build GRCh38
rs377152124 Chr.2: 219174735-219174735 on Build GRCh38

View Full Product Details