Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13681791-13682062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM210A
Entrez Gene ID
125228
Gene Name
family with sequence similarity 210 member A
Gene Aliases
C18orf19, HsT2329
UniGene
Hs.13034
Gene Chromosome Location
Chr.18: 13663347-13726592 on Build GRCh38
Transcripts
NM_001098801.1 NM_152352.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6500315 c.18A>G p.P6P Chr.18: 13682060-13682060 on Build GRCh38
6298290 c.227C>A p.P76H Chr.18: 13681851-13681851 on Build GRCh38
1189651 c.149G>C p.G50A Chr.18: 13681929-13681929 on Build GRCh38
5666502 c.276C>A p.F92L Chr.18: 13681802-13681802 on Build GRCh38
3422068 c.157A>C p.K53Q Chr.18: 13681921-13681921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767849715 Chr.18: 13681936-13681936 on Build GRCh38
rs552796973 Chr.18: 13682038-13682038 on Build GRCh38
rs758549366 Chr.18: 13682030-13682030 on Build GRCh38
rs145075680 Chr.18: 13681877-13681877 on Build GRCh38
rs377543711 Chr.18: 13681875-13681875 on Build GRCh38
rs764167668 Chr.18: 13681918-13681918 on Build GRCh38
rs201619883 Chr.18: 13681952-13681952 on Build GRCh38
rs780936180 Chr.18: 13681999-13681999 on Build GRCh38
rs759972433 Chr.18: 13681844-13681844 on Build GRCh38
rs143865357 Chr.18: 13681900-13681900 on Build GRCh38
rs763468795 Chr.18: 13682005-13682005 on Build GRCh38
rs748077437 Chr.18: 13681896-13681896 on Build GRCh38
rs373346025 Chr.18: 13681848-13681848 on Build GRCh38
rs779670880 Chr.18: 13682036-13682037 on Build GRCh38
rs777433452 Chr.18: 13681981-13681981 on Build GRCh38
rs753137291 Chr.18: 13681852-13681852 on Build GRCh38
rs767475426 Chr.18: 13682021-13682021 on Build GRCh38
rs774144774 Chr.18: 13681818-13681818 on Build GRCh38
rs760768896 Chr.18: 13682037-13682037 on Build GRCh38
rs745483549 Chr.18: 13682002-13682002 on Build GRCh38
rs375228347 Chr.18: 13682027-13682027 on Build GRCh38
rs771730636 Chr.18: 13681897-13681897 on Build GRCh38
rs113115642 Chr.18: 13681990-13681990 on Build GRCh38
rs771679397 Chr.18: 13681831-13681831 on Build GRCh38
rs369772010 Chr.18: 13681842-13681842 on Build GRCh38
rs35493157 Chr.18: 13681963-13681963 on Build GRCh38
rs779729816 Chr.18: 13681876-13681876 on Build GRCh38
rs189782979 Chr.18: 13681915-13681915 on Build GRCh38
rs751584591 Chr.18: 13681922-13681922 on Build GRCh38
rs986155289 Chr.18: 13681917-13681917 on Build GRCh38
rs757273055 Chr.18: 13681930-13681930 on Build GRCh38
rs747729503 Chr.18: 13681826-13681826 on Build GRCh38
rs758344817 Chr.18: 13681967-13681967 on Build GRCh38
rs201878744 Chr.18: 13681867-13681867 on Build GRCh38
rs752482708 Chr.18: 13682041-13682041 on Build GRCh38
rs552403778 Chr.18: 13682012-13682012 on Build GRCh38
rs151157304 Chr.18: 13681911-13681911 on Build GRCh38
rs766683697 Chr.18: 13681834-13681834 on Build GRCh38
rs754852546 Chr.18: 13681959-13681959 on Build GRCh38
rs878933292 Chr.18: 13681929-13681929 on Build GRCh38
rs148603152 Chr.18: 13681902-13681902 on Build GRCh38
rs116967198 Chr.18: 13681833-13681833 on Build GRCh38
rs769212391 Chr.18: 13682010-13682010 on Build GRCh38
rs200885719 Chr.18: 13681884-13681884 on Build GRCh38
rs770339833 Chr.18: 13681998-13681998 on Build GRCh38
rs754726735 Chr.18: 13681880-13681880 on Build GRCh38
rs144093105 Chr.18: 13681956-13681956 on Build GRCh38
rs776132977 Chr.18: 13681910-13681910 on Build GRCh38
rs577321430 Chr.18: 13681868-13681868 on Build GRCh38
rs775768207 Chr.18: 13682004-13682004 on Build GRCh38
rs932727900 Chr.18: 13681969-13681969 on Build GRCh38
rs142125636 Chr.18: 13681904-13681904 on Build GRCh38
rs765484785 Chr.18: 13681913-13681913 on Build GRCh38
rs548721066 Chr.18: 13681912-13681912 on Build GRCh38
rs770213503 Chr.18: 13682003-13682003 on Build GRCh38
rs758510315 Chr.18: 13681855-13681855 on Build GRCh38
rs761995037 Chr.18: 13682019-13682019 on Build GRCh38
rs140347489 Chr.18: 13681916-13681916 on Build GRCh38
rs770557083 Chr.18: 13681908-13681908 on Build GRCh38
rs746574984 Chr.18: 13681995-13681995 on Build GRCh38

View Full Product Details