Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64300270-64300525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK4-TEX40
Entrez Gene ID
106780802
Gene Name
KCNK4-TEX40 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.11: 64291722-64304769 on Build GRCh38
Transcripts
NR_133662.1 NM_001039496.1

Gene Symbol
TEX40
Entrez Gene ID
25858
Gene Name
testis expressed 40
Gene Aliases
C11orf20
UniGene
Hs.97174
Gene Chromosome Location
Chr.11: 64300391-64304767 on Build GRCh38
Transcripts
NR_133662.1 NM_001039496.1

SNP Detail

SNP ID Chromosome Location
rs202061149 Chr.11: 64300397-64300397 on Build GRCh38
rs756750237 Chr.11: 64300447-64300447 on Build GRCh38
rs1039479773 Chr.11: 64300376-64300376 on Build GRCh38
rs748977209 Chr.11: 64300463-64300463 on Build GRCh38
rs911665626 Chr.11: 64300493-64300493 on Build GRCh38
rs377274910 Chr.11: 64300439-64300439 on Build GRCh38
rs762862526 Chr.11: 64300375-64300375 on Build GRCh38
rs376331366 Chr.11: 64300428-64300428 on Build GRCh38
rs375983366 Chr.11: 64300410-64300410 on Build GRCh38
rs779883369 Chr.11: 64300466-64300466 on Build GRCh38
rs3741402 Chr.11: 64300308-64300308 on Build GRCh38
rs1052025561 Chr.11: 64300340-64300340 on Build GRCh38
rs45572341 Chr.11: 64300417-64300417 on Build GRCh38
rs1007808209 Chr.11: 64300349-64300349 on Build GRCh38
rs751266914 Chr.11: 64300445-64300445 on Build GRCh38
rs929430560 Chr.11: 64300337-64300337 on Build GRCh38
rs990929252 Chr.11: 64300297-64300297 on Build GRCh38
rs749056605 Chr.11: 64300471-64300471 on Build GRCh38
rs781774577 Chr.11: 64300343-64300343 on Build GRCh38
rs761175423 Chr.11: 64300413-64300413 on Build GRCh38
rs559272264 Chr.11: 64300321-64300321 on Build GRCh38
rs749593553 Chr.11: 64300438-64300446 on Build GRCh38
rs535283432 Chr.11: 64300389-64300389 on Build GRCh38
rs368198219 Chr.11: 64300490-64300490 on Build GRCh38
rs941135357 Chr.11: 64300322-64300322 on Build GRCh38
rs774753414 Chr.11: 64300380-64300380 on Build GRCh38
rs746352592 Chr.11: 64300379-64300379 on Build GRCh38
rs745876751 Chr.11: 64300370-64300370 on Build GRCh38
rs369586049 Chr.11: 64300374-64300374 on Build GRCh38
rs767762768 Chr.11: 64300393-64300393 on Build GRCh38
rs745376047 Chr.11: 64300461-64300461 on Build GRCh38
rs959070538 Chr.11: 64300295-64300295 on Build GRCh38
rs374713474 Chr.11: 64300458-64300458 on Build GRCh38
rs761961510 Chr.11: 64300381-64300381 on Build GRCh38
rs754182637 Chr.11: 64300423-64300423 on Build GRCh38
rs144624337 Chr.11: 64300328-64300328 on Build GRCh38
rs572117296 Chr.11: 64300324-64300324 on Build GRCh38
rs372758856 Chr.11: 64300398-64300398 on Build GRCh38
rs987598831 Chr.11: 64300491-64300492 on Build GRCh38
rs756027576 Chr.11: 64300464-64300464 on Build GRCh38

View Full Product Details