Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50660129-50660390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIP2B
Entrez Gene ID
57609
Gene Name
disco interacting protein 2 homolog B
Gene Aliases
-
UniGene
Hs.505516
Gene Chromosome Location
Chr.12: 50504985-50748667 on Build GRCh38
Transcripts
NM_173602.2 XM_005269044.1 XM_006719520.3 XM_006719521.3 XM_011538594.2 XM_011538595.2 XM_011538596.2 XM_017019699.1 XM_017019700.1 XM_017019701.1 XM_017019702.1 XM_017019703.1 XM_017019704.1 XM_017019705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4779416 c.343A>G p.K115E Chr.12: 50660235-50660235 on Build GRCh38
3671095 c.366C>A p.P122P Chr.12: 50660258-50660258 on Build GRCh38
4042602 c.358G>A p.A120T Chr.12: 50660250-50660250 on Build GRCh38
6462671 c.374C>A p.T125N Chr.12: 50660266-50660266 on Build GRCh38
1135393 c.396T>C p.V132V Chr.12: 50660288-50660288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765617618 Chr.12: 50660161-50660161 on Build GRCh38
rs1032784088 Chr.12: 50660265-50660265 on Build GRCh38
rs751068226 Chr.12: 50660195-50660195 on Build GRCh38
rs367778126 Chr.12: 50660247-50660247 on Build GRCh38
rs762660129 Chr.12: 50660173-50660173 on Build GRCh38
rs746377467 Chr.12: 50660339-50660339 on Build GRCh38
rs78788593 Chr.12: 50660182-50660182 on Build GRCh38
rs1019702513 Chr.12: 50660316-50660316 on Build GRCh38
rs528654977 Chr.12: 50660160-50660160 on Build GRCh38
rs750391754 Chr.12: 50660205-50660205 on Build GRCh38
rs767295582 Chr.12: 50660202-50660202 on Build GRCh38
rs771404002 Chr.12: 50660313-50660315 on Build GRCh38
rs770241507 Chr.12: 50660351-50660351 on Build GRCh38
rs571235285 Chr.12: 50660197-50660197 on Build GRCh38
rs965419576 Chr.12: 50660323-50660323 on Build GRCh38
rs1037221879 Chr.12: 50660315-50660315 on Build GRCh38
rs575430146 Chr.12: 50660328-50660328 on Build GRCh38
rs148198953 Chr.12: 50660303-50660303 on Build GRCh38
rs777198568 Chr.12: 50660159-50660159 on Build GRCh38
rs200447869 Chr.12: 50660359-50660359 on Build GRCh38
rs779894888 Chr.12: 50660238-50660238 on Build GRCh38
rs747025155 Chr.12: 50660297-50660297 on Build GRCh38
rs904454228 Chr.12: 50660247-50660247 on Build GRCh38
rs776059881 Chr.12: 50660164-50660164 on Build GRCh38
rs774896601 Chr.12: 50660340-50660341 on Build GRCh38
rs17124837 Chr.12: 50660153-50660153 on Build GRCh38
rs201078128 Chr.12: 50660222-50660222 on Build GRCh38
rs557247973 Chr.12: 50660163-50660163 on Build GRCh38
rs758565805 Chr.12: 50660259-50660259 on Build GRCh38
rs897311547 Chr.12: 50660342-50660342 on Build GRCh38
rs771300456 Chr.12: 50660156-50660156 on Build GRCh38

View Full Product Details