Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 64552197-64552444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFTPH
Entrez Gene ID
54812
Gene Name
aftiphilin
Gene Aliases
Nbla10388
UniGene
Hs.655167
Gene Chromosome Location
Chr.2: 64524305-64593005 on Build GRCh38
Transcripts
NM_001002243.2 NM_017657.4 NM_203437.3 XM_005264380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4095135 c.752C>T p.A251V Chr.2: 64552226-64552226 on Build GRCh38
4609774 c.793_795delAAT p.N265delN Chr.2: 64552267-64552269 on Build GRCh38
5445031 c.739G>C p.E247Q Chr.2: 64552213-64552213 on Build GRCh38
4095137 c.785G>A p.R262Q Chr.2: 64552259-64552259 on Build GRCh38
5726962 c.745G>A p.E249K Chr.2: 64552219-64552219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768079005 Chr.2: 64552361-64552361 on Build GRCh38
rs892837332 Chr.2: 64552272-64552272 on Build GRCh38
rs762201238 Chr.2: 64552278-64552278 on Build GRCh38
rs138916712 Chr.2: 64552384-64552384 on Build GRCh38
rs766564086 Chr.2: 64552308-64552308 on Build GRCh38
rs918291 Chr.2: 64552265-64552265 on Build GRCh38
rs767781919 Chr.2: 64552279-64552279 on Build GRCh38
rs982314118 Chr.2: 64552357-64552357 on Build GRCh38
rs752949972 Chr.2: 64552418-64552418 on Build GRCh38
rs766176038 Chr.2: 64552324-64552324 on Build GRCh38
rs755134312 Chr.2: 64552312-64552312 on Build GRCh38
rs530739373 Chr.2: 64552382-64552382 on Build GRCh38
rs775045835 Chr.2: 64552259-64552259 on Build GRCh38
rs754011963 Chr.2: 64552310-64552310 on Build GRCh38
rs774956181 Chr.2: 64552345-64552345 on Build GRCh38
rs146936440 Chr.2: 64552366-64552366 on Build GRCh38
rs765244594 Chr.2: 64552405-64552405 on Build GRCh38
rs761390354 Chr.2: 64552369-64552369 on Build GRCh38
rs77781733 Chr.2: 64552337-64552337 on Build GRCh38
rs565051653 Chr.2: 64552378-64552378 on Build GRCh38
rs181965139 Chr.2: 64552284-64552284 on Build GRCh38
rs754418577 Chr.2: 64552400-64552400 on Build GRCh38
rs555921915 Chr.2: 64552380-64552380 on Build GRCh38
rs150198863 Chr.2: 64552248-64552248 on Build GRCh38
rs370251115 Chr.2: 64552315-64552315 on Build GRCh38
rs201111661 Chr.2: 64552275-64552275 on Build GRCh38
rs138062318 Chr.2: 64552318-64552318 on Build GRCh38
rs762990474 Chr.2: 64552291-64552291 on Build GRCh38
rs750906119 Chr.2: 64552290-64552290 on Build GRCh38
rs769760598 Chr.2: 64552344-64552344 on Build GRCh38
rs549069405 Chr.2: 64552303-64552303 on Build GRCh38
rs868773184 Chr.2: 64552261-64552261 on Build GRCh38
rs781020750 Chr.2: 64552332-64552332 on Build GRCh38
rs758613179 Chr.2: 64552420-64552420 on Build GRCh38
rs3770740 Chr.2: 64552375-64552375 on Build GRCh38
rs763853003 Chr.2: 64552268-64552268 on Build GRCh38
rs770654545 Chr.2: 64552245-64552245 on Build GRCh38
rs757018577 Chr.2: 64552319-64552319 on Build GRCh38
rs780932733 Chr.2: 64552422-64552422 on Build GRCh38
rs748742884 Chr.2: 64552346-64552346 on Build GRCh38
rs746871941 Chr.2: 64552237-64552237 on Build GRCh38
rs1025960424 Chr.2: 64552235-64552235 on Build GRCh38
rs200609046 Chr.2: 64552258-64552258 on Build GRCh38
rs375002466 Chr.2: 64552401-64552401 on Build GRCh38

View Full Product Details